540242
(R)-(−)-1,2-Propanediol
96%
Synonym(s):
(R)-(−)-Propylene glycerol, (R)-(−)-Propylene glycol
Sign Into View Organizational & Contract Pricing
All Photos(1)
About This Item
Linear Formula:
CH3CH(OH)CH2OH
CAS Number:
Molecular Weight:
76.09
Beilstein:
1718872
MDL number:
UNSPSC Code:
12352001
PubChem Substance ID:
NACRES:
NA.22
Recommended Products
Assay
96%
optical activity
[α]20/D −16.5°, neat
optical purity
ee: 98% (GLC)
refractive index
n20/D 1.434 (lit.)
bp
186-188 °C/765 mmHg (lit.)
density
1.04 g/mL at 25 °C (lit.)
functional group
hydroxyl
SMILES string
C[C@@H](O)CO
InChI
1S/C3H8O2/c1-3(5)2-4/h3-5H,2H2,1H3/t3-/m1/s1
InChI key
DNIAPMSPPWPWGF-GSVOUGTGSA-N
Looking for similar products? Visit Product Comparison Guide
Other Notes
(R)-(-)-1,2-Propanediol undergoes metabolization in the bovine rumen fluid by the rumen microbiota with the emission of sulfur-containing gases.
Legal Information
Manufactured under license by Sterling Pharma Solutions Limited, using Jacobsen HKR technology.
Storage Class Code
10 - Combustible liquids
WGK
WGK 3
Flash Point(F)
217.4 °F - closed cup
Flash Point(C)
103 °C - closed cup
Personal Protective Equipment
dust mask type N95 (US), Eyeshields, Gloves
Choose from one of the most recent versions:
Already Own This Product?
Find documentation for the products that you have recently purchased in the Document Library.
Customers Also Viewed
Yang Zhao et al.
Nature communications, 8, 14180-14180 (2017-01-26)
Many naturally occurring biomolecules, such as amino acids, sugars and nucleotides, are inherently chiral. Enantiomers, a pair of chiral isomers with opposite handedness, often exhibit similar physical and chemical properties due to their identical functional groups and composition, yet show
Ruminal fermentation of propylene glycol and glycerol.
Trabue S, et al.
Journal of Agricultural and Food Chemistry, 55(17), 7043-7051 (2007)
Propylene glycol and glycerol concentration in ultrasound gel.
David Belavy et al.
Regional anesthesia and pain medicine, 38(1), 75-76 (2012-12-25)
Rong-Yao Wang et al.
Physical chemistry chemical physics : PCCP, 15(9), 3313-3319 (2013-01-31)
Understanding the role of kinetics in fiber network microstructure formation is of considerable importance in engineering gel materials to achieve their optimized performances/functionalities. In this work, we present a new approach for kinetic-structure analysis for fibrous gel materials. In this
R Mateus et al.
International journal of pharmaceutics, 444(1-2), 106-108 (2013-01-30)
Confocal Raman spectroscopy (CRS) is a novel approach which has been used to determine stratum corneum thickness and to profile endogenous skin components. However, the utility of this technique for probing drug disposition in vivo has not been explored to
Chromatograms
suitable for GCOur team of scientists has experience in all areas of research including Life Science, Material Science, Chemical Synthesis, Chromatography, Analytical and many others.
Contact Technical Service