Skip to Content
Merck
All Photos(1)

Key Documents

538507

Sigma-Aldrich

2-Amino-6-bromobenzothiazole

97%

Sign Into View Organizational & Contract Pricing


About This Item

Empirical Formula (Hill Notation):
C7H5BrN2S
CAS Number:
Molecular Weight:
229.10
MDL number:
UNSPSC Code:
12352100
PubChem Substance ID:
NACRES:
NA.22

Assay

97%

form

solid

mp

213-217 °C (lit.)

functional group

bromo

SMILES string

Nc1nc2ccc(Br)cc2s1

InChI

1S/C7H5BrN2S/c8-4-1-2-5-6(3-4)11-7(9)10-5/h1-3H,(H2,9,10)

InChI key

VZEBSJIOUMDNLY-UHFFFAOYSA-N

Looking for similar products? Visit Product Comparison Guide

General description

Reaction of 4-bromoaniline with acetic acid and potassium thiocynate in solution of bromine in acetic acid yields 2-amino-6-bromobenzothiazole. 2-Amino-6-bromobenzothiazole can also be synthesized from 2-aminobenzothiazole via monobromination with silica-supported quinolinium tribromide.

Application

2-Amino-6-bromobenzothiazole may be used in the synthesis of the following
  • 2-amino-6-arylbenzothiazole compounds:
  • 6-p-tolylbenzo[d]thiazole-2-amine
  • 6-(4chlorophenyl)benzo[d]thiazole-2-amine
  • 6-(4-methoxyphenyl)benzo[d]thiazole-2-amine
  • 6-(3,5-bis(triflouromethyl)phenyl)benzo[d]thiazole-2-amine
  • 6-phenylbenzo[d]thiazole-2-amine
It may be employed as a ligand (L) in the synthesis of organometallic derivatives of Rh(I) [Rh(CO)2(L)(Cl)].

Pictograms

Exclamation mark

Signal Word

Warning

Hazard Statements

Hazard Classifications

Acute Tox. 4 Oral - Eye Irrit. 2 - Skin Sens. 1

Storage Class Code

11 - Combustible Solids

WGK

WGK 3

Flash Point(F)

Not applicable

Flash Point(C)

Not applicable

Personal Protective Equipment

dust mask type N95 (US), Eyeshields, Gloves

Choose from one of the most recent versions:

Certificates of Analysis (COA)

Lot/Batch Number

Don't see the Right Version?

If you require a particular version, you can look up a specific certificate by the Lot or Batch number.

Already Own This Product?

Find documentation for the products that you have recently purchased in the Document Library.

Visit the Document Library

Silica-supported quinolinium tribromide: a recoverable solid brominating reagent for regioselective monobromination of aromatic amines.
Li Z, et al.
Journal of the Brazilian Chemical Society, 21(3), 496-501 (2010)
Efficient synthesis of 2-amino-6-arylbenzothiazoles via Pd (0) Suzuki cross coupling reactions: potent urease enzyme inhibition and nitric oxide scavenging activities of the products.
Gull Y, et al.
Molecules (Basel), 18(8), 8845-8857 (2013)
Synthesis, cytostatic, and antitumor properties of new Rh (I) thiazole complexes.
Craciunescu, D. G., et al.
Biological trace element research, 8(4), 251-261 (1985)
Claire Bagnéris et al.
Proceedings of the National Academy of Sciences of the United States of America, 111(23), 8428-8433 (2014-05-23)
Voltage-gated sodium channels are important targets for the development of pharmaceutical drugs, because mutations in different human sodium channel isoforms have causal relationships with a range of neurological and cardiovascular diseases. In this study, functional electrophysiological studies show that the

Our team of scientists has experience in all areas of research including Life Science, Material Science, Chemical Synthesis, Chromatography, Analytical and many others.

Contact Technical Service