534463
1,3,5,7,9,11,14-Heptaisobutyltricyclo[7.3.3.15,11]heptasiloxane-endo-3,7,14-triol
97%
Synonym(s):
Isobutyltrisilanol-POSS®
Sign Into View Organizational & Contract Pricing
All Photos(1)
About This Item
Empirical Formula (Hill Notation):
C28H66O12Si7
CAS Number:
Molecular Weight:
791.41
MDL number:
UNSPSC Code:
12162002
PubChem Substance ID:
NACRES:
NA.23
Recommended Products
Assay
97%
mp
139-143 °C (lit.)
SMILES string
CC(C)C[Si]1(O)O[Si]2(CC(C)C)O[Si](O)(CC(C)C)O[Si]3(CC(C)C)O[Si](O)(CC(C)C)O[Si](CC(C)C)(O1)O[Si](CC(C)C)(O2)O3
InChI
1S/C28H66O12Si7/c1-22(2)15-41(29)32-44(18-25(7)8)34-42(30,16-23(3)4)36-46(20-27(11)12)37-43(31,17-24(5)6)35-45(33-41,19-26(9)10)39-47(38-44,40-46)21-28(13)14/h22-31H,15-21H2,1-14H3
InChI key
APIBTMSFBUJAAC-UHFFFAOYSA-N
Legal Information
POSS is a registered trademark of Hybrid Plastics, Inc.
Storage Class Code
11 - Combustible Solids
WGK
WGK 3
Flash Point(F)
Not applicable
Flash Point(C)
Not applicable
Personal Protective Equipment
dust mask type N95 (US), Eyeshields, Gloves
Choose from one of the most recent versions:
Already Own This Product?
Find documentation for the products that you have recently purchased in the Document Library.
Customers Also Viewed
Our team of scientists has experience in all areas of research including Life Science, Material Science, Chemical Synthesis, Chromatography, Analytical and many others.
Contact Technical Service