Skip to Content
Merck
All Photos(2)

Key Documents

534080

Sigma-Aldrich

3-Bromophenethylamine

97%

Sign Into View Organizational & Contract Pricing


About This Item

Linear Formula:
BrC6H4(CH2)2NH2
CAS Number:
Molecular Weight:
200.08
MDL number:
UNSPSC Code:
12352100
PubChem Substance ID:
NACRES:
NA.22

Quality Level

Assay

97%

refractive index

n20/D 1.5740 (lit.)

bp

239-240 °C (lit.)

density

1.406 g/mL at 25 °C (lit.)

SMILES string

NCCc1cccc(Br)c1

InChI

1S/C8H10BrN/c9-8-3-1-2-7(6-8)4-5-10/h1-3,6H,4-5,10H2

InChI key

ORHRHMLEFQBHND-UHFFFAOYSA-N

Pictograms

Corrosion

Signal Word

Danger

Hazard Statements

Hazard Classifications

Eye Dam. 1 - Skin Corr. 1B

Storage Class Code

8A - Combustible corrosive hazardous materials

WGK

WGK 3

Flash Point(F)

>230.0 °F - closed cup

Flash Point(C)

> 110 °C - closed cup

Personal Protective Equipment

dust mask type N95 (US), Eyeshields, Gloves

Certificates of Analysis (COA)

Search for Certificates of Analysis (COA) by entering the products Lot/Batch Number. Lot and Batch Numbers can be found on a product’s label following the words ‘Lot’ or ‘Batch’.

Already Own This Product?

Find documentation for the products that you have recently purchased in the Document Library.

Visit the Document Library

Máté Erdélyi et al.
Chemistry (Weinheim an der Bergstrasse, Germany), 12(2), 403-412 (2005-09-28)
Peptide secondary structure mimetics are important tools in medicinal chemistry, as they provide analogues of endogenous peptides with new physicochemical and pharmacological properties. The development, synthesis, photochemical investigation, and conformational analysis of a stilbene-type beta-hairpin mimetic capable of light-triggered conformational
Matthias M Herth et al.
Bioorganic & medicinal chemistry, 20(14), 4574-4581 (2012-06-12)
2-(2',6'-Dimethoxy-[1,1'-biphenyl]-3-yl)-N,N-dimethylethanamine has been identified as a potent ligand for the serotonin 7 (5-HT(7)) receptor. In this study, we describe the synthesis, radiolabeling and in vivo evaluation of [(11)C]2-(2',6'-dimethoxy-[1,1'-biphenyl]-3-yl)-N,N-dimethylethanamine ([(11)C]Cimbi-806) as a radioligand for imaging brain 5-HT(7) receptors with positron emission
Magalie Paillet-Loilier et al.
Bioorganic & medicinal chemistry letters, 17(11), 3018-3022 (2007-04-10)
The synthesis of a series of aminoethylbiphenyls as novel 5-HT(7) receptor ligands is described. The novel derivatives exhibit high affinity for the 5-HT(7) receptor with selectivity toward 5-HT(1A) receptor.

Our team of scientists has experience in all areas of research including Life Science, Material Science, Chemical Synthesis, Chromatography, Analytical and many others.

Contact Technical Service