Skip to Content
Merck
All Photos(1)

Key Documents

533874

Sigma-Aldrich

Bromoiodomethane

97%

Sign Into View Organizational & Contract Pricing


About This Item

Linear Formula:
BrCH2I
CAS Number:
Molecular Weight:
220.84
MDL number:
UNSPSC Code:
12352100
PubChem Substance ID:
NACRES:
NA.22

Quality Level

Assay

97%

contains

copper as stabilizer

refractive index

n20/D 1.6382 (lit.)

bp

138-141 °C (lit.)

density

2.93 g/mL at 25 °C (lit.)

functional group

bromo
iodo

SMILES string

BrCI

InChI

1S/CH2BrI/c2-1-3/h1H2

InChI key

TUDWMIUPYRKEFN-UHFFFAOYSA-N

Looking for similar products? Visit Product Comparison Guide

Related Categories

Application

Reactant for:
  • Carbene insertion reactions
  • Ultrafast photodissociation

Pictograms

CorrosionExclamation mark

Signal Word

Danger

Hazard Statements

Hazard Classifications

Eye Dam. 1 - Skin Irrit. 2 - STOT SE 3

Target Organs

Respiratory system

Storage Class Code

10 - Combustible liquids

WGK

WGK 3

Flash Point(F)

Not applicable

Flash Point(C)

Not applicable

Personal Protective Equipment

dust mask type N95 (US), Eyeshields, Gloves

Choose from one of the most recent versions:

Certificates of Analysis (COA)

Lot/Batch Number

Don't see the Right Version?

If you require a particular version, you can look up a specific certificate by the Lot or Batch number.

Already Own This Product?

Find documentation for the products that you have recently purchased in the Document Library.

Visit the Document Library

Nicholas J Lodge et al.
Nuclear medicine and biology, 41(6), 524-535 (2014-05-06)
A radioligand for measuring the density of corticotropin-releasing factor subtype-1 receptors (CRF1 receptors) in living animal and human brain with positron emission tomography (PET) would be a useful tool for neuropsychiatric investigations and the development of drugs intended to interact

Our team of scientists has experience in all areas of research including Life Science, Material Science, Chemical Synthesis, Chromatography, Analytical and many others.

Contact Technical Service