Skip to Content
Merck
All Photos(1)

Key Documents

532169

Sigma-Aldrich

(R)-(−)-N-Boc-3-pyrrolidinol

98%

Synonym(s):

(R)-N-tert-Butoxycarbonyl-(−)-3-pyrrolidinol

Sign Into View Organizational & Contract Pricing


About This Item

Empirical Formula (Hill Notation):
C9H17NO3
CAS Number:
Molecular Weight:
187.24
MDL number:
UNSPSC Code:
12352005
PubChem Substance ID:
NACRES:
NA.22

Quality Level

Assay

98%

optical activity

[α]20/D −26±1, c = 1 in methanol

mp

62-65 °C (lit.)

functional group

hydroxyl

SMILES string

CC(C)(C)OC(=O)N1CC[C@@H](O)C1

InChI

1S/C9H17NO3/c1-9(2,3)13-8(12)10-5-4-7(11)6-10/h7,11H,4-6H2,1-3H3/t7-/m1/s1

InChI key

APCBTRDHCDOPNY-SSDOTTSWSA-N

Application

Used in synthesis of ligands for the nicotinic acetylcholine receptor.
Used in the synthesis of anticoagulants.

Pictograms

Exclamation markEnvironment

Signal Word

Warning

Hazard Statements

Hazard Classifications

Aquatic Acute 1 - Eye Irrit. 2 - Skin Irrit. 2 - STOT SE 3

Target Organs

Respiratory system

Storage Class Code

11 - Combustible Solids

WGK

WGK 3

Personal Protective Equipment

dust mask type N95 (US), Eyeshields, Gloves

Choose from one of the most recent versions:

Certificates of Analysis (COA)

Lot/Batch Number

Don't see the Right Version?

If you require a particular version, you can look up a specific certificate by the Lot or Batch number.

Already Own This Product?

Find documentation for the products that you have recently purchased in the Document Library.

Visit the Document Library

J Lee et al.
Bioorganic & medicinal chemistry letters, 10(10), 1063-1066 (2000-06-08)
The preparation of novel pyridyl ethers as ligands for the nicotinic acetylcholine receptor (nAChR) is described. Variations of the ring size of the azacycle and substitution on the pyridine had dramatic effects on receptor binding affinity with IC50s at the
Satoshi Komoriya et al.
Bioorganic & medicinal chemistry, 12(9), 2099-2114 (2004-04-15)
Since factor Xa (fXa) plays a pivotal role in the blood coagulation cascade, inhibition of fXa is thought to be an effective treatment for a variety of thrombotic events. (2S)-2-[4-[[(3S)-1-Acetimidoyl-3-pyrrolidinyl]oxy]phenyl]-3-(7-amidino-2-naphthyl)propanoic acid hydrochloride pentahydrate (DX-9065a) was previously found in our laboratory

Our team of scientists has experience in all areas of research including Life Science, Material Science, Chemical Synthesis, Chromatography, Analytical and many others.

Contact Technical Service