Skip to Content
Merck
All Photos(1)

Key Documents

527424

Sigma-Aldrich

2-Hydroxy-3-nitrobenzaldehyde

98%

Synonym(s):

3-Nitrosalicylaldehyde

Sign Into View Organizational & Contract Pricing


About This Item

Linear Formula:
HOC6H3(NO2)CHO
CAS Number:
Molecular Weight:
167.12
EC Number:
MDL number:
UNSPSC Code:
12352100
PubChem Substance ID:
NACRES:
NA.22

Quality Level

Assay

98%

mp

105-109 °C (lit.)

SMILES string

Oc1c(C=O)cccc1[N+]([O-])=O

InChI

1S/C7H5NO4/c9-4-5-2-1-3-6(7(5)10)8(11)12/h1-4,10H

InChI key

NUGOTBXFVWXVTE-UHFFFAOYSA-N

General description

2-Hydroxy-3-nitrobenzaldehyde can be extracted from the leaves of Actephila merrilliana. Its molecule is planar. On reaction with dapsone, it affords a colorless solution.

Application

2-Hydroxy-3-nitrobenzaldehyde may be used in the synthesis of 5-acetyl-4-aryl-6-methyl-1,2,3,4-tetrahydro pyrimidine.

Pictograms

Exclamation mark

Signal Word

Warning

Hazard Statements

Hazard Classifications

Eye Irrit. 2 - Skin Irrit. 2 - STOT SE 3

Target Organs

Respiratory system

Storage Class Code

11 - Combustible Solids

WGK

WGK 3

Flash Point(F)

Not applicable

Flash Point(C)

Not applicable

Personal Protective Equipment

dust mask type N95 (US), Eyeshields, Gloves

Certificates of Analysis (COA)

Search for Certificates of Analysis (COA) by entering the products Lot/Batch Number. Lot and Batch Numbers can be found on a product’s label following the words ‘Lot’ or ‘Batch’.

Already Own This Product?

Find documentation for the products that you have recently purchased in the Document Library.

Visit the Document Library

Customers Also Viewed

Synthesis of new 4-aryl-isoxazolo [5, 4-d] pyrimidin-6-one (thione) and 4-aryl-pyrazolo [3, 4-d]-pyrimidin-6-one derivatives of potential antihypertensive activity.
El-Hamouly WS, et al.
Indian J. Chem. B, 45(9), 2091-2091 (2006)
2-Hydroxy-3-nitrobenzaldehyde.
Tang B, et al.
Acta Crystallographica Section E, Structure Reports Online, 66(8), o1912-o1912 (2010)
Dapsone assay based on Schiff base formation.
Levy L and Higgins LJ.
Blood, 4(2) (1966)

Our team of scientists has experience in all areas of research including Life Science, Material Science, Chemical Synthesis, Chromatography, Analytical and many others.

Contact Technical Service