Skip to Content
Merck
All Photos(3)

Key Documents

523151

Sigma-Aldrich

4-Bromo-3-nitrobenzaldehyde

97%

Sign Into View Organizational & Contract Pricing


About This Item

Linear Formula:
BrC6H3(NO2)CHO
CAS Number:
Molecular Weight:
230.02
MDL number:
UNSPSC Code:
12352100
PubChem Substance ID:
NACRES:
NA.22

Assay

97%

form

solid

mp

104-108 °C (lit.)

functional group

aldehyde
bromo
nitro

SMILES string

[O-][N+](=O)c1cc(C=O)ccc1Br

InChI

1S/C7H4BrNO3/c8-6-2-1-5(4-10)3-7(6)9(11)12/h1-4H

InChI key

SAFSVELFSYQXOV-UHFFFAOYSA-N

Pictograms

Exclamation markEnvironment

Signal Word

Warning

Hazard Statements

Hazard Classifications

Aquatic Acute 1 - Eye Irrit. 2 - Skin Irrit. 2 - Skin Sens. 1 - STOT SE 3

Target Organs

Respiratory system

Storage Class Code

11 - Combustible Solids

WGK

WGK 3

Flash Point(F)

Not applicable

Flash Point(C)

Not applicable

Personal Protective Equipment

dust mask type N95 (US), Eyeshields, Gloves

Choose from one of the most recent versions:

Certificates of Analysis (COA)

Lot/Batch Number

Don't see the Right Version?

If you require a particular version, you can look up a specific certificate by the Lot or Batch number.

Already Own This Product?

Find documentation for the products that you have recently purchased in the Document Library.

Visit the Document Library

Jian-Bo Wang et al.
Chemical communications (Cambridge, England), 48(5), 744-746 (2011-11-29)
A novel fluorescent probe for gold ions (Au(3+)/Au(+)) is reported through blocking photoinduced electron transfer, in which a boron dipyrromethene (Bodipy) derivative reveals high selectivity and sensitivity in a gold-catalyzed intramolecular hydroamination, and is successfully applied to fluorescence imaging of
Synthesis and characterization of a conjugated polymer with stable radicals in the side groups.
Swoboda P, et al.
Macromolecules, 28(12), 4255-4259 (1995)
Chunyan Du et al.
The Journal of organic chemistry, 74(19), 7322-7327 (2009-08-29)
Two asymmetrically substituted dicyanovinyl heterotetracenes are synthesized and characterized. Single-crystal X-ray diffractions indicate both intermolecular hydrogen bonds and pi-stacking existing in the solid-state structures. The solvatochromic behaviors of the two heterotetracenes were investigated in a variety of solvents. Both experimental

Our team of scientists has experience in all areas of research including Life Science, Material Science, Chemical Synthesis, Chromatography, Analytical and many others.

Contact Technical Service