Skip to Content
Merck
All Photos(3)

Key Documents

52200

Sigma-Aldrich

Hexacosanoic acid

technical, ≥90% (GC)

Synonym(s):

Cerotic acid, Cerotinic acid

Sign Into View Organizational & Contract Pricing


About This Item

Linear Formula:
CH3(CH2)24COOH
CAS Number:
Molecular Weight:
396.69
Beilstein:
1799681
EC Number:
MDL number:
UNSPSC Code:
12352100
PubChem Substance ID:
NACRES:
NA.22

grade

technical

Assay

≥90% (GC)

mp

84-88 °C

functional group

carboxylic acid

SMILES string

CCCCCCCCCCCCCCCCCCCCCCCCCC(O)=O

InChI

1S/C26H52O2/c1-2-3-4-5-6-7-8-9-10-11-12-13-14-15-16-17-18-19-20-21-22-23-24-25-26(27)28/h2-25H2,1H3,(H,27,28)

InChI key

XMHIUKTWLZUKEX-UHFFFAOYSA-N

Looking for similar products? Visit Product Comparison Guide

Storage Class Code

11 - Combustible Solids

WGK

WGK 3

Flash Point(F)

Not applicable

Flash Point(C)

Not applicable

Personal Protective Equipment

dust mask type N95 (US), Eyeshields, Gloves

Choose from one of the most recent versions:

Certificates of Analysis (COA)

Lot/Batch Number

Don't see the Right Version?

If you require a particular version, you can look up a specific certificate by the Lot or Batch number.

Already Own This Product?

Find documentation for the products that you have recently purchased in the Document Library.

Visit the Document Library

Customers Also Viewed

Slide 1 of 2

1 of 2

Pedro Brites et al.
Brain : a journal of neurology, 132(Pt 2), 482-492 (2008-11-22)
Peroxisomes are organelles responsible for multiple metabolic pathways including, the biosynthesis of plasmalogens, a class of phospholipids, and the beta-oxidation of very-long-chain fatty acids (VLCFA). Lack of peroxisomes or dysfunction in any of their normal functions is the cellular basis
P Aubourg et al.
The New England journal of medicine, 329(11), 745-752 (1993-09-09)
Adrenomyeloneuropathy is an X-linked recessive disorder characterized by myelopathy, peripheral neuropathy, and cerebral demyelination, which develop in association with the accumulation of very-long-chain fatty acids. The administration of oleic and erucic acids inhibits the synthesis of very-long-chain fatty acids. Recently
Marc Engelen et al.
Journal of inherited metabolic disease, 35(6), 1137-1145 (2012-03-27)
X-linked adrenoleukodystrophy (X-ALD) is caused by mutations in the ABCD1 gene encoding ALDP, an ATP-binding-cassette (ABC) transporter located in the peroxisomal membrane. ALDP deficiency results in impaired peroxisomal β-oxidation and the subsequent accumulation of very long-chain fatty acids (VLCFA; >
Hugo W Moser et al.
Archives of neurology, 62(7), 1073-1080 (2005-07-13)
To identify asymptomatic boys with X-linked adrenoleukodystrophy who have a normal magnetic resonance image (MRI), and to assess the effect of 4:1 glyceryl trioleate-glyceryl trierucate (Lorenzo's oil) on disease progression. Eighty-nine boys (mean +/- SD baseline age, 4.7 +/- 4.1
A Sadeghi-Nejad et al.
The New England journal of medicine, 322(1), 13-16 (1990-01-04)
Adrenoleukodystrophy, a sex-linked peroxisomal disorder that results in the impaired oxidation of long-chain saturated fatty acids and causes neurologic impairment, is a rare cause of Addison's disease in children. Adrenomyeloneuropathy is the name given to a biochemically identical but milder

Our team of scientists has experience in all areas of research including Life Science, Material Science, Chemical Synthesis, Chromatography, Analytical and many others.

Contact Technical Service