Skip to Content
Merck
All Photos(2)

Key Documents

520039

Sigma-Aldrich

2-Ethynylbenzyl alcohol

95%

Sign Into View Organizational & Contract Pricing


About This Item

Linear Formula:
HC≡CC6H4CH2OH
CAS Number:
Molecular Weight:
132.16
MDL number:
UNSPSC Code:
12352100
PubChem Substance ID:
NACRES:
NA.22

Assay

95%

form

solid

mp

66-70 °C (lit.)

SMILES string

OCc1ccccc1C#C

InChI

1S/C9H8O/c1-2-8-5-3-4-6-9(8)7-10/h1,3-6,10H,7H2

InChI key

ZKCWIJQOZHADEM-UHFFFAOYSA-N

Related Categories

Pictograms

Health hazardExclamation mark

Signal Word

Danger

Hazard Statements

Hazard Classifications

Acute Tox. 4 Oral - Eye Irrit. 2 - Resp. Sens. 1

Storage Class Code

11 - Combustible Solids

WGK

WGK 2

Personal Protective Equipment

dust mask type N95 (US), Eyeshields, Gloves

Certificates of Analysis (COA)

Search for Certificates of Analysis (COA) by entering the products Lot/Batch Number. Lot and Batch Numbers can be found on a product’s label following the words ‘Lot’ or ‘Batch’.

Already Own This Product?

Find documentation for the products that you have recently purchased in the Document Library.

Visit the Document Library

Preparation, Structural and Spectral Properties of Poly (2-ethynylbenzyl alcohol).
Lee WC, et al.
Mol. Cryst. Liq. Cryst., 513(1), 196-204 (2009)
Cyclization reactions of 2-alkynylbenzyl alcohol and 2-alkynylbenzylamine derivatives promoted by tetrabutylammonium fluoride.
Hiroya K, et al.
Tetrahedron, 57(48), 9697-9710 (2001)
Synthesis of 1-(Alkoxycarbonyl) methylene-1, 3-dihydroisobenzofurans and 4-(Alkoxycarbonyl) benzo [c] pyrans by Palladium-Catalysed Oxidative Carbonylation of 2-Alkynylbenzyl Alcohols, 2-Alkynylbenzaldehydes and 2-Alkynylphenyl Ketones.
Bacchi A, et al.
European Journal of Organic Chemistry, 3, 574-585 (2004)
Xueqing Wang et al.
The Journal of organic chemistry, 63(21), 7357-7363 (2001-10-24)
An attempt to trap radical intermediates during the monoamine oxidase (MAO) catalyzed oxidation of amines by intramolecular cyclization with an activated alkene that is built into the substrate was unsuccessful. (E)-2-(Iodoethenyl)benzylamine (3a) was shown to be a reversible inhibitor of
Subhasish Tapadar et al.
Bioorganic & medicinal chemistry, 23(24), 7543-7564 (2015-11-21)
Inhibition of the enzymatic activity of histone deacetylase (HDAC) is a promising therapeutic strategy for cancer treatment and several distinct small molecule histone deacetylase inhibitors (HDACi) have been reported. We have previously identified a new class of non-peptide macrocyclic HDACi

Articles

Alkynes' versatility enables reactions like addition, metathesis, hydroboration, cleavage, coupling, and cycloadditions in synthetic chemistry.

Alkynes' versatility enables reactions like addition, metathesis, hydroboration, cleavage, coupling, and cycloadditions in synthetic chemistry.

Alkynes' versatility enables reactions like addition, metathesis, hydroboration, cleavage, coupling, and cycloadditions in synthetic chemistry.

Alkynes' versatility enables reactions like addition, metathesis, hydroboration, cleavage, coupling, and cycloadditions in synthetic chemistry.

Our team of scientists has experience in all areas of research including Life Science, Material Science, Chemical Synthesis, Chromatography, Analytical and many others.

Contact Technical Service