Skip to Content
Merck

498297

Sigma-Aldrich

4,6-Diaminoresorcinol dihydrochloride

97%

Sign Into View Organizational & Contract Pricing


About This Item

Linear Formula:
(H2N)2C6H2-1,3-(OH)2 · 2HCl
CAS Number:
Molecular Weight:
213.06
MDL number:
UNSPSC Code:
12352100
PubChem Substance ID:
NACRES:
NA.22

Pricing and availability is not currently available.

Quality Level

Assay

97%

mp

254 °C (dec.) (lit.)

storage temp.

2-8°C

SMILES string

Cl.Cl.Nc1cc(N)c(O)cc1O

InChI

1S/C6H8N2O2.2ClH/c7-3-1-4(8)6(10)2-5(3)9;;/h1-2,9-10H,7-8H2;2*1H

InChI key

KUMOYHHELWKOCB-UHFFFAOYSA-N

Compare Similar Items

View Full Comparison

Show Differences

1 of 4

This Item
B22852665525193755
storage temp.

2-8°C

storage temp.

-

storage temp.

2-8°C

storage temp.

-

mp

254 °C (dec.) (lit.)

mp

143-145 °C (dec.) (lit.)

mp

173-177 °C

mp

264 °C (dec.) (lit.)

Application

4,6-Diaminoresorcinol dihydrochloride may be used as a precursor in the synthesis of:
  • poly(p-phenylene benzobisoxazole) (PBO)[1][2]
  • poly(2,6-naphthalenebenzobisoxazole) (Naph-2,6-PBO)[3]
  • poly(1,5-naphthalenebenzobisoxazole) (Naph-1,5-PBO)[3]

Storage Class Code

11 - Combustible Solids

WGK

WGK 3

Flash Point(F)

Not applicable

Flash Point(C)

Not applicable

Personal Protective Equipment

dust mask type N95 (US), Eyeshields, Gloves

Choose from one of the most recent versions:

Certificates of Analysis (COA)

Lot/Batch Number

Don't see the Right Version?

If you require a particular version, you can look up a specific certificate by the Lot or Batch number.

Already Own This Product?

Find documentation for the products that you have recently purchased in the Document Library.

Visit the Document Library

Synthesis, structure and properties of carbon nanotube/poly(p-phenylene benzobisoxazole) composite fibres.
Li J, et al.
Polymer International, 55(4), 456-465 (2006)
Facile sulfur-assisted carbonylation of diaminoresorcinol with carbon monoxide.
Mizuno T, et al.
Heteroatom Chem., 23(1), 111-116 (2012)
A safe cost-efficient synthesis of 4,6-diaminoresorcinol.
Pews RG, et al.
The Journal of Organic Chemistry, 62(23), 8255-8256 (1997)
The synthesis, characterization, and crystal structures of poly (2,6-naphthalenebenzobisoxazole) and poly (1,5-naphthalenebenzobisoxazole).
Park SY, et al.
Journal of Polymer Science. Part B, Polymer Physics, 44(14), 1948-1957 (2006)

Our team of scientists has experience in all areas of research including Life Science, Material Science, Chemical Synthesis, Chromatography, Analytical and many others.

Contact Technical Service