Skip to Content
Merck
All Photos(1)

Key Documents

498262

Sigma-Aldrich

Diphenyl selenide

96%

Sign Into View Organizational & Contract Pricing


About This Item

Linear Formula:
(C6H5)2Se
CAS Number:
Molecular Weight:
233.17
EC Number:
MDL number:
UNSPSC Code:
12352100
PubChem Substance ID:
NACRES:
NA.22

Quality Level

Assay

96%

refractive index

n20/D 1.6465 (lit.)

bp

115-117 °C (lit.)

density

1.338 g/mL at 25 °C (lit.)

storage temp.

2-8°C

SMILES string

[Se](c1ccccc1)c2ccccc2

InChI

1S/C12H10Se/c1-3-7-11(8-4-1)13-12-9-5-2-6-10-12/h1-10H

InChI key

ORQWTLCYLDRDHK-UHFFFAOYSA-N

General description

Diphenyl selenide is an organoselenium compound. Its standard enthalpies of combustion, formation and the mean bond-dissociation energy have been calculated. Its potential as a neutral carrier to develop silver-selective membrane electrode has been investigated. Diphenyl selenide undergoes oxidation to form selenoxides on treating with N-bromosuccinimide followed by alkaline hydrolysis.

Application

Used for Anti-selective Michael addition of thiols and their analogs to nitro-olefins.

Signal Word

Danger

Hazard Statements

Hazard Classifications

Acute Tox. 3 Inhalation - Acute Tox. 3 Oral - Aquatic Acute 1 - Aquatic Chronic 1 - STOT RE 2

Storage Class Code

6.1C - Combustible acute toxic Cat.3 / toxic compounds or compounds which causing chronic effects

WGK

WGK 3

Flash Point(F)

>230.0 °F - closed cup

Flash Point(C)

> 110 °C - closed cup

Personal Protective Equipment

dust mask type N95 (US), Eyeshields, Gloves

Choose from one of the most recent versions:

Certificates of Analysis (COA)

Lot/Batch Number

Don't see the Right Version?

If you require a particular version, you can look up a specific certificate by the Lot or Batch number.

Already Own This Product?

Find documentation for the products that you have recently purchased in the Document Library.

Visit the Document Library

Organoselenide as a novel ionophore for a silver-selective membrane electrode.
Katsu T and Xu D.
Analytical Letters, 31(12), 1979-1989 (1998)
Enthalpies of combustion of selenium and diphenyl selenide.
Barnes DS and Mortimer CT.
The Journal of Chemical Thermodynamics, 5(3), 371-377 (1973)
The Journal of Organic Chemistry, 55, 2437-2437 (1990)
Oxidation of selenides and tellurides with positive halogenating species.
Detty MR
The Journal of Organic Chemistry, 45(2), 274-279 (1980)
Xinghua Dong et al.
ACS nano, 14(5), 5400-5416 (2020-04-24)
Radiotherapy (RT) in practical use often suffers from off-target side effects and ineffectiveness against hypoxic tumor microenvironment (TME) as well as remote metastases. With regard to these problems, herein, we provide semiconductor heterojunction structured WO2.9-WSe2-PEG nanoparticles to realize a synergistic

Our team of scientists has experience in all areas of research including Life Science, Material Science, Chemical Synthesis, Chromatography, Analytical and many others.

Contact Technical Service