Recommended Products
isotopic purity
98 atom % 15N
Quality Level
form
solid
mp
232-234 °C (lit.)
mass shift
M+1
SMILES string
Cl.C[15NH2]
InChI
1S/CH5N.ClH/c1-2;/h2H2,1H3;1H/i2+1;
InChI key
NQMRYBIKMRVZLB-CGOMOMTCSA-N
Related Categories
Packaging
This product may be available from bulk stock and can be packaged on demand. For information on pricing, availability and packaging, please contact Stable Isotopes Customer Service.
Signal Word
Warning
Hazard Statements
Hazard Classifications
Acute Tox. 4 Oral
Storage Class Code
11 - Combustible Solids
WGK
WGK 3
Flash Point(F)
Not applicable
Flash Point(C)
Not applicable
Personal Protective Equipment
dust mask type N95 (US), Eyeshields, Gloves
Choose from one of the most recent versions:
Already Own This Product?
Find documentation for the products that you have recently purchased in the Document Library.
Molecular bioSystems, 8(11), 2891-2900 (2012-08-01)
Molecular dynamics simulations have been performed to investigate the binding of tris(hydroxymethyl)-aminomethane to the surface of the core domain of the mouse cellular tumor antigen p53 employing the GROMOS and 53A6 force field parameter sets. A close investigation of the
Physical chemistry chemical physics : PCCP, 15(13), 4707-4714 (2013-02-21)
Using density functional theory calculations, we report on the adsorption of methylamine on gold and compare its adsorption to a selection of alkylamines, methanol and methanethiol. On the (111) surface, the amines, thiol and alcohol bind in the ontop site
Microbial ecology, 65(1), 180-196 (2012-09-04)
Ammonia is the preferred nitrogen source for many algae including the cyanobacterium Synechococcus elongatis (Synechococcus R-2; PCC 7942). Modelling ammonia uptake by cells is not straightforward because it exists in solution as NH(3) and NH (4) (+) . NH(3) is
Journal of ethnopharmacology, 142(3), 647-656 (2012-06-13)
Zhibai Dihuang Pill (ZDP) is one of ancient traditional Chinese medicines (TCMs), which is usually used for the treatment of kidney deficiency for thousands of years in China. Traditional Chinese medicines (TCMs) usually operate in vivo through multi-components, multi-ways and
Journal of bacteriology, 195(10), 2359-2367 (2013-03-19)
The methylotroph Methylobacterium extorquens AM1 oxidizes methanol and methylamine to formaldehyde and subsequently to formate, an intermediate that serves as the branch point between assimilation (formation of biomass) and dissimilation (oxidation to CO₂). The oxidation of formaldehyde to formate is
Our team of scientists has experience in all areas of research including Life Science, Material Science, Chemical Synthesis, Chromatography, Analytical and many others.
Contact Technical Service