489255
Ethane-d6
99 atom % D, gas
Synonym(s):
Hexadeuteroethane
Sign Into View Organizational & Contract Pricing
All Photos(1)
About This Item
Linear Formula:
CD3CD3
CAS Number:
Molecular Weight:
36.11
MDL number:
UNSPSC Code:
12142200
PubChem Substance ID:
NACRES:
NA.12
Recommended Products
isotopic purity
99 atom % D
Assay
99% (CP)
form
gas
bp
−88 °C (lit.)
mp
−172 °C (lit.)
mass shift
M+6
SMILES string
[2H]C([2H])([2H])C([2H])([2H])[2H]
InChI
1S/C2H6/c1-2/h1-2H3/i1D3,2D3
InChI key
OTMSDBZUPAUEDD-WFGJKAKNSA-N
Packaging
Standard 1 L quantity is packaged in a 450 mL carbon steel lecture bottle with brass CGA 110/180 valve. Nominal gas pressure at 21ºC is 20 psig for 1 L. This pressure is slightly above atmospheric pressure. Care must be taken when extracting this product from the cylinder.
This product may be available from bulk stock and can be packaged on demand. For information on pricing, availability and packaging, please contact Stable Isotopes Customer Service.
Other Notes
We are in the process of phasing-out the CGA 110/170 valves in favor of CGA 110/180 valves. Some stock of cylinders with CGA 110/170 valves may still exist. Please use adapters Z147095-1EA (brass) or Z147125-1EA (stainless steel) to utilize with a CGA 180 regulator.
Recommended products
Stainless steel control valves Z146951 or Z187224
PTFE sealing washers GCA inlet size 180 Z146994
PTFE sealing washers CGA inlet size 110 Z146986
PTFE sealing washers GCA inlet size 180 Z146994
PTFE sealing washers CGA inlet size 110 Z146986
Signal Word
Danger
Hazard Statements
Precautionary Statements
Hazard Classifications
Flam. Gas 1A - Press. Gas Compr. Gas
Storage Class Code
2A - Gases
WGK
nwg
Flash Point(F)
-211.0 °F - closed cup
Flash Point(C)
-135 °C - closed cup
Choose from one of the most recent versions:
Already Own This Product?
Find documentation for the products that you have recently purchased in the Document Library.
Our team of scientists has experience in all areas of research including Life Science, Material Science, Chemical Synthesis, Chromatography, Analytical and many others.
Contact Technical Service