488666
Decanoic-d19 acid
98 atom % D, 98% (CP)
Synonym(s):
Capric acid-d19,OH
About This Item
Recommended Products
isotopic purity
98 atom % D
Quality Level
Assay
98% (CP)
form
solid
bp
268-270 °C (lit.)
mp
30-32 °C (lit.)
density
0.991 g/mL at 25 °C
mass shift
M+19
SMILES string
[2H]C([2H])([2H])C([2H])([2H])C([2H])([2H])C([2H])([2H])C([2H])([2H])C([2H])([2H])C([2H])([2H])C([2H])([2H])C([2H])([2H])C(O)=O
InChI
1S/C10H20O2/c1-2-3-4-5-6-7-8-9-10(11)12/h2-9H2,1H3,(H,11,12)/i1D3,2D2,3D2,4D2,5D2,6D2,7D2,8D2,9D2
InChI key
GHVNFZFCNZKVNT-KIJKOTCYSA-N
Related Categories
Packaging
Signal Word
Warning
Hazard Statements
Precautionary Statements
Hazard Classifications
Aquatic Chronic 3 - Eye Irrit. 2 - Skin Irrit. 2
Storage Class Code
11 - Combustible Solids
WGK
WGK 1
Flash Point(F)
230.0 °F - closed cup
Flash Point(C)
110.00 °C - closed cup
Choose from one of the most recent versions:
Already Own This Product?
Find documentation for the products that you have recently purchased in the Document Library.
Our team of scientists has experience in all areas of research including Life Science, Material Science, Chemical Synthesis, Chromatography, Analytical and many others.
Contact Technical Service