All Photos(1)
About This Item
Linear Formula:
HOCH2CH2N(CH3)213CH3Br
CAS Number:
Molecular Weight:
185.07
MDL number:
UNSPSC Code:
12352106
PubChem Substance ID:
NACRES:
NA.12
Recommended Products
isotopic purity
99 atom % 13C
form
solid
technique(s)
mass spectrometry (MS): suitable
mass shift
M+1
SMILES string
[Br-].C[N+](C)([13CH3])CCO
InChI
1S/C5H14NO.BrH/c1-6(2,3)4-5-7;/h7H,4-5H2,1-3H3;1H/q+1;/p-1/i1+1;
InChI key
JJCWKVUUIFLXNZ-YTBWXGASSA-M
Packaging
This product may be available from bulk stock and can be packaged on demand. For information on pricing, availability and packaging, please contact Stable Isotopes Customer Service.
Storage Class Code
11 - Combustible Solids
WGK
WGK 3
Flash Point(F)
Not applicable
Flash Point(C)
Not applicable
Choose from one of the most recent versions:
Already Own This Product?
Find documentation for the products that you have recently purchased in the Document Library.
Our team of scientists has experience in all areas of research including Life Science, Material Science, Chemical Synthesis, Chromatography, Analytical and many others.
Contact Technical Service