Skip to Content
Merck
All Photos(1)

Key Documents

487791

Sigma-Aldrich

Acetic acid-18O2

95 atom % 18O

Sign Into View Organizational & Contract Pricing


About This Item

Linear Formula:
CH3C18O2H
CAS Number:
Molecular Weight:
64.05
MDL number:
UNSPSC Code:
12352106
PubChem Substance ID:
NACRES:
NA.12

isotopic purity

95 atom % 18O

Quality Level

refractive index

n20/D 1.3715 (lit.)

bp

117-118 °C (lit.)

mp

16.2 °C (lit.)

density

1.116 g/mL at 25 °C

mass shift

M+4

SMILES string

CC([18OH])=[18O]

InChI

1S/C2H4O2/c1-2(3)4/h1H3,(H,3,4)/i3+2,4+2

InChI key

QTBSBXVTEAMEQO-NUQCWPJISA-N

Packaging

This product may be available from bulk stock and can be packaged on demand. For information on pricing, availability and packaging, please contact Stable Isotopes Customer Service.

Pictograms

FlameCorrosion

Signal Word

Danger

Hazard Statements

Hazard Classifications

Eye Dam. 1 - Flam. Liq. 3 - Skin Corr. 1A

Storage Class Code

3 - Flammable liquids

WGK

WGK 1

Flash Point(F)

104.0 °F - closed cup

Flash Point(C)

40 °C - closed cup

Personal Protective Equipment

dust mask type N95 (US), Eyeshields, Gloves

Choose from one of the most recent versions:

Certificates of Analysis (COA)

Lot/Batch Number

Don't see the Right Version?

If you require a particular version, you can look up a specific certificate by the Lot or Batch number.

Already Own This Product?

Find documentation for the products that you have recently purchased in the Document Library.

Visit the Document Library

Customers Also Viewed

Our team of scientists has experience in all areas of research including Life Science, Material Science, Chemical Synthesis, Chromatography, Analytical and many others.

Contact Technical Service