Skip to Content
Merck
All Photos(1)

Key Documents

48700

Sigma-Aldrich

Octyl gallate

antioxidant, ≥99.0% (HPLC)

Synonym(s):

Octyl 3,4,5-trihydroxybenzoate, n-Octyl gallate

Sign Into View Organizational & Contract Pricing


About This Item

Linear Formula:
3,4,5-(HO)3C6H2CO2(CH2)7CH3
CAS Number:
Molecular Weight:
282.33
Beilstein:
2132305
EC Number:
MDL number:
UNSPSC Code:
12352100
PubChem Substance ID:
NACRES:
NA.22

Assay

≥99.0% (HPLC)

mp

101-103 °C
101-104 °C (lit.)

solubility

soluble 2.5%, clear, colorless (AcOH (MEOH))

functional group

ester

SMILES string

CCCCCCCCOC(=O)c1cc(O)c(O)c(O)c1

InChI

1S/C15H22O5/c1-2-3-4-5-6-7-8-20-15(19)11-9-12(16)14(18)13(17)10-11/h9-10,16-18H,2-8H2,1H3

InChI key

NRPKURNSADTHLJ-UHFFFAOYSA-N

Looking for similar products? Visit Product Comparison Guide

General description

Octyl gallate (3,4,5-trihydroxybenzoate) is reported to possess antifungal activity against Saccharomyces cerevisiae and Zygosaccharomyces bailii. The fungicidal activity associated with octyl gallate is due to its ability to act as a nonionic surface-active agent (surfactant). Octyl gallate, an alkyl gallate, is reported to exhibit direct antibacterial activity against Staphylococcus aureus BB568. It is reported to inhibit the activity of soybean lipoxygenase-1 (EC 1.13.11.12, type I) with an IC50 of 1.3μM. It is also effective in preventing lipid peroxidation.

Application

Octyl gallate was used in a study to investigate the self-assembly in two comb-shaped supramolecules systems consisting of octyl gallate, hydrogen bonded to the pyridine groups of polyisoprene-block-poly(vinylpyridine) diblock copolymers using synchrotron radiation.

Disclaimer

The product is not intended for use as a biocide under global biocide regulations, including but not limited to US EPA′s Federal Insecticide Fungicide and Rodenticide Act, European Biocidal Products Regulation, Canada’s Pest Management Regulatory Agency, Turkey’s Biocidal Products Regulation, Korea’s Consumer Chemical Products and Biocide Safety Management Act (K-BPR) and others.

Pictograms

Exclamation mark

Signal Word

Warning

Hazard Statements

Hazard Classifications

Acute Tox. 4 Oral - Skin Sens. 1

Storage Class Code

11 - Combustible Solids

WGK

WGK 1

Flash Point(F)

Not applicable

Flash Point(C)

Not applicable

Personal Protective Equipment

dust mask type N95 (US), Eyeshields, Gloves

Choose from one of the most recent versions:

Certificates of Analysis (COA)

Lot/Batch Number

Don't see the Right Version?

If you require a particular version, you can look up a specific certificate by the Lot or Batch number.

Already Own This Product?

Find documentation for the products that you have recently purchased in the Document Library.

Visit the Document Library

Self-Assembly of Supramolecules Consisting of Octyl Gallate Hydrogen Bonded to Polyisoprene-b lock-poly (vinylpyridine) Diblock Copolymers.
Bondzic S, et al.
Macromolecules, 37(25), 9517-9524 (2004)
Fu-Lan Hsu et al.
Bioresource technology, 98(4), 734-738 (2006-06-06)
The objective of this study was to investigate the possibility of using octyl gallate alone or with organic biocides as a preservative against wood decay fungi. Antifungal activities of three antioxidants, propyl gallate, octyl gallate and butylated hydroxyltoluene (BHT) were
E Sierra-Campos et al.
Microbiology (Reading, England), 155(Pt 2), 604-611 (2009-02-10)
The effects of octyl gallate on Ustilago maydis yeast cells were analysed in relation to its capacity to oxidize compounds (pro-oxidant actions). All phenolic compounds tested inhibited the alternative oxidase (AOX). However, only octyl gallate induced a morphological change in
Eve-Marie Josse et al.
European journal of biochemistry, 270(18), 3787-3794 (2003-09-03)
The plastid terminal oxidase (PTOX) encoded by the Arabidopsis IMMUTANS gene was expressed in Escherichia coli cells and its quinone/oxygen oxidoreductase activity monitored in isolated bacterial membranes using NADH as an electron donor. Specificity for plastoquinone was observed. Neither ubiquinone
Liisa Törmäkangas et al.
Biochemical pharmacology, 70(8), 1222-1230 (2005-09-06)
Increasing evidence suggests that plant polyphenolic compounds may protect from cardiovascular diseases, which have been addressed to their antioxidative properties. In addition, these compounds have been shown to possess anti-inflammatory and anti-microbial potential. In the present study we tested the

Our team of scientists has experience in all areas of research including Life Science, Material Science, Chemical Synthesis, Chromatography, Analytical and many others.

Contact Technical Service