484652
6-Bromopyridine-2-carboxylic acid
98%
Synonym(s):
6-Bromopicolinic acid
Sign Into View Organizational & Contract Pricing
All Photos(2)
About This Item
Empirical Formula (Hill Notation):
C6H4BrNO2
CAS Number:
Molecular Weight:
202.01
MDL number:
UNSPSC Code:
12352100
PubChem Substance ID:
NACRES:
NA.22
Recommended Products
Assay
98%
mp
192-194 °C (lit.)
functional group
bromo
carboxylic acid
SMILES string
OC(=O)c1cccc(Br)n1
InChI
1S/C6H4BrNO2/c7-5-3-1-2-4(8-5)6(9)10/h1-3H,(H,9,10)
InChI key
XURXQNUIGWHWHU-UHFFFAOYSA-N
Related Categories
Application
Used in synthesis of chiral 2,2′-dipyidylamines
Signal Word
Warning
Hazard Statements
Precautionary Statements
Hazard Classifications
Eye Irrit. 2 - Skin Irrit. 2 - STOT SE 3
Target Organs
Respiratory system
Storage Class Code
11 - Combustible Solids
WGK
WGK 3
Flash Point(F)
Not applicable
Flash Point(C)
Not applicable
Personal Protective Equipment
dust mask type N95 (US), Eyeshields, Gloves
Choose from one of the most recent versions:
Already Own This Product?
Find documentation for the products that you have recently purchased in the Document Library.
Tetrahedron Letters, 45, 5019-5021 (2004)
Our team of scientists has experience in all areas of research including Life Science, Material Science, Chemical Synthesis, Chromatography, Analytical and many others.
Contact Technical Service