Skip to Content
Merck
All Photos(1)

Key Documents

484318

Sigma-Aldrich

Ethylene glycol monophenyl ether

≥90%

Synonym(s):

DOWANOL® EPh, 2-Phenoxyethanol, Phenylglycol

Sign Into View Organizational & Contract Pricing


About This Item

Linear Formula:
C6H5OCH2CH2OH
CAS Number:
Molecular Weight:
138.16
Beilstein:
1364011
EC Number:
MDL number:
UNSPSC Code:
12162002
PubChem Substance ID:
NACRES:
NA.23

vapor density

4.8 (vs air)

Quality Level

vapor pressure

0.01 mmHg ( 20 °C)

Assay

≥90%

form

liquid

impurities

≤0.5% Phenol
<10% diethylene glycol phenyl ether

refractive index

n20/D 1.537 (lit.)

pH

7 (23 °C, 10 g/L)

bp

247 °C (lit.)

mp

11-13 °C (lit.)

density

1.102 g/mL at 25 °C (lit.)

SMILES string

OCCOc1ccccc1

InChI

1S/C8H10O2/c9-6-7-10-8-4-2-1-3-5-8/h1-5,9H,6-7H2

InChI key

QCDWFXQBSFUVSP-UHFFFAOYSA-N

Looking for similar products? Visit Product Comparison Guide

Legal Information

Product of Dow Chemical
™Trademark of The Dow Chemical Company
DOWANOL is a registered trademark of The Dow Chemical Company or an affiliated company of Dow

Disclaimer

The product is not intended for use as a biocide under global biocide regulations, including but not limited to US EPA′s Federal Insecticide Fungicide and Rodenticide Act, European Biocidal Products Regulation, Canada’s Pest Management Regulatory Agency, Turkey’s Biocidal Products Regulation, Korea’s Consumer Chemical Products and Biocide Safety Management Act (K-BPR) and others.

Pictograms

CorrosionExclamation mark

Signal Word

Danger

Hazard Statements

Hazard Classifications

Acute Tox. 4 Oral - Eye Dam. 1 - STOT SE 3

Target Organs

Respiratory system

Storage Class Code

10 - Combustible liquids

WGK

WGK 1

Flash Point(F)

258.8 °F - closed cup

Flash Point(C)

126 °C - closed cup

Personal Protective Equipment

dust mask type N95 (US), Eyeshields, Gloves

Certificates of Analysis (COA)

Search for Certificates of Analysis (COA) by entering the products Lot/Batch Number. Lot and Batch Numbers can be found on a product’s label following the words ‘Lot’ or ‘Batch’.

Already Own This Product?

Find documentation for the products that you have recently purchased in the Document Library.

Visit the Document Library

Yi Shen et al.
Environmental science & technology, 49(12), 7364-7372 (2015-05-27)
Graphene nanosheets, as a novel nanoadsorbent, can be further modified to optimize the adsorption capability for various pollutants. To overcome the structural limits of graphene (aggregation) and graphene oxide (hydrophilic surface) in water, sulfonated graphene (GS) was prepared by diazotization
Therese Lloyd et al.
BMC medicine, 13, 171-171 (2015-08-01)
In the UK, a man's lifetime risk of being diagnosed with prostate cancer is 1 in 8. We calculated both the lifetime risk of being diagnosed with and dying from prostate cancer by major ethnic group. Public Health England provided
Joshua Quick et al.
Genome biology, 16, 114-114 (2015-05-31)
Foodborne outbreaks of Salmonella remain a pressing public health concern. We recently detected a large outbreak of Salmonella enterica serovar Enteritidis phage type 14b affecting more than 30 patients in our hospital. This outbreak was linked to community, national and
Shao-Ping Weng et al.
Reproductive toxicology (Elmsford, N.Y.), 29(4), 452-457 (2010-04-13)
Ethylene glycol monomethyl ether (EGME) is known to alter the reproductive function of exposed animals and their offspring; however, its influence on cumulus-oocyte complexes (COCs) remains unclear. This study assessed the toxicity of EGME on oocytes and COCs by analyzing
J Lademann et al.
Skin pharmacology and physiology, 25(2), 100-106 (2012-02-04)
Colonization and infection of wounds represent a major reason for the impairment of tissue repair. Recently, it has been reported that tissue-tolerable plasma (TTP) is highly efficient in the reduction of the bacterial load of the skin. In the present

Our team of scientists has experience in all areas of research including Life Science, Material Science, Chemical Synthesis, Chromatography, Analytical and many others.

Contact Technical Service