Skip to Content
Merck
All Photos(1)

Key Documents

483443

Sigma-Aldrich

Tetradecyl isocyanate

97%

Synonym(s):

1-Isocyanatotetradecane, Myristyl isocyanate, n-Tetradecyl isocyanate

Sign Into View Organizational & Contract Pricing


About This Item

Linear Formula:
CH3(CH2)13NCO
CAS Number:
Molecular Weight:
239.40
MDL number:
UNSPSC Code:
12352100
PubChem Substance ID:
NACRES:
NA.22

Quality Level

Assay

97%

refractive index

n20/D 1.446 (lit.)

bp

165-170 °C/1.4 mmHg (lit.)

density

0.869 g/mL at 25 °C (lit.)

functional group

amine
isocyanate

storage temp.

2-8°C

SMILES string

CCCCCCCCCCCCCCN=C=O

InChI

1S/C15H29NO/c1-2-3-4-5-6-7-8-9-10-11-12-13-14-16-15-17/h2-14H2,1H3

InChI key

CSMJMAQKBKGDQX-UHFFFAOYSA-N

Related Categories

Application

  • Synthesis of hydrophobically modified ethoxylated non-isocyanate urethanes (HENIURs) and their use as rheology additives: Details the application of Tetradecyl isocyanate in the production of rheology additives, relevant for material science, especially in enhancing the properties of various materials (B Quienne et al., 2022).

Pictograms

Health hazardExclamation mark

Signal Word

Danger

Hazard Classifications

Acute Tox. 4 Dermal - Acute Tox. 4 Inhalation - Acute Tox. 4 Oral - Eye Irrit. 2 - Resp. Sens. 1 - Skin Irrit. 2 - Skin Sens. 1 - STOT SE 3

Target Organs

Respiratory system

Storage Class Code

6.1C - Combustible acute toxic Cat.3 / toxic compounds or compounds which causing chronic effects

WGK

WGK 3

Flash Point(F)

235.4 °F - closed cup

Flash Point(C)

113 °C - closed cup

Personal Protective Equipment

dust mask type N95 (US), Eyeshields, Gloves

Choose from one of the most recent versions:

Certificates of Analysis (COA)

Lot/Batch Number

Don't see the Right Version?

If you require a particular version, you can look up a specific certificate by the Lot or Batch number.

Already Own This Product?

Find documentation for the products that you have recently purchased in the Document Library.

Visit the Document Library

Our team of scientists has experience in all areas of research including Life Science, Material Science, Chemical Synthesis, Chromatography, Analytical and many others.

Contact Technical Service