Skip to Content
Merck
All Photos(4)

Key Documents

483036

Sigma-Aldrich

Fullerene-C60

greener alternative

98%

Synonym(s):

Buckminsterfullerene

Sign Into View Organizational & Contract Pricing


About This Item

Empirical Formula (Hill Notation):
C60
CAS Number:
Molecular Weight:
720.64
Beilstein:
5901022
MDL number:
UNSPSC Code:
12352103
NACRES:
NA.23

Assay

98%

form

powder

greener alternative product characteristics

Design for Energy Efficiency
Learn more about the Principles of Green Chemistry.

sustainability

Greener Alternative Product

mp

>280 °C (lit.)

solubility

organic solvents: soluble
organic solvents: soluble

Orbital energy

HOMO 6.1 eV 
LUMO 4.5 eV 

OLED Device Performance

ITO/CuPc/NPD/Alq3/C60/Mg:Ag

  • Color: green
  • Max. Luminance: 17200 Cd/m2

ITO/NPD/Alq3/C60/LiF:Al:Ag
  • Color: green
  • Max. Luminance: 7000 Cd/m2
  • Turn-On Voltage: <5 V

ITO/TPD/Alq3/C60/LiF/Al/Ag
  • Color: green
  • Max. Luminance: 4000 Cd/m2
  • Turn-On Voltage: <5 V

greener alternative category

SMILES string

c12c3c4c5c1c6c7c8c2c9c%10c3c%11c%12c4c%13c%14c5c%15c6c%16c7c%17c%18c8c9c%19c%20c%10c%11c%21c%22c%12c%13c%23c%24c%14c%15c%25c%16c%26c%17c%27c%18c%19c%28c%20c%21c%29c%22c%23c%30c%24c%25c%26c%31c%27c%28c%29c%30%31

InChI

1S/C60/c1-2-5-6-3(1)8-12-10-4(1)9-11-7(2)17-21-13(5)23-24-14(6)22-18(8)28-20(12)30-26-16(10)15(9)25-29-19(11)27(17)37-41-31(21)33(23)43-44-34(24)32(22)42-38(28)48-40(30)46-36(26)35(25)45-39(29)47(37)55-49(41)51(43)57-52(44)50(42)56(48)59-54(46)53(45)58(55)60(57)59

InChI key

XMWRBQBLMFGWIX-UHFFFAOYSA-N

Looking for similar products? Visit Product Comparison Guide

General description

Fullerene-C60 (C60) is an allotrope of carbon that has a closed-cage structure and it is icosahedrally shaped with 60 sp2 hybridized carbons. It is majorly utilized in the developement of novel materials for a wide range of energy based applications.
We are committed to bringing you Greener Alternative Products, which adhere to one or more of The 12 Principles of Greener Chemistry. This product has been enhanced for energy efficiency. Find details here.

Application

C60 is a nanomaterial that can be used in the fabrication of a variety of electrochemical devices which include solar cells, lithium ion batteries and photovoltaic cells.
Fullerene-C60 (98%) belongs to the class of carbonaceous materials. It can be used for a variety of alternative energy applications, which include lithium-ion batteries, organic photovoltaic cells (OPV), heterojunction solar cells, polymeric photodetectors, photodiodes, and photocatalysis.

Pictograms

Exclamation mark

Signal Word

Warning

Hazard Statements

Hazard Classifications

Eye Irrit. 2 - STOT SE 3

Target Organs

Respiratory system

Storage Class Code

11 - Combustible Solids

WGK

WGK 3

Flash Point(F)

Not applicable

Flash Point(C)

Not applicable

Personal Protective Equipment

dust mask type N95 (US), Eyeshields, Gloves

Choose from one of the most recent versions:

Certificates of Analysis (COA)

Lot/Batch Number

Don't see the Right Version?

If you require a particular version, you can look up a specific certificate by the Lot or Batch number.

Already Own This Product?

Find documentation for the products that you have recently purchased in the Document Library.

Visit the Document Library

Oscillatory shear rheology of polystyrene melts filled with carbon black and fullerene
Plastics, Rubber and Composites, 41(9), 384-389 (2012)
The Effect of Colloidal n-C60 on DNA Mutagenesis and the SOS DNA Repair Mechanism of Salmonella typhimurium strains TA1538 and TA1535.
Chu S, et al.
Journal of Experimental Microbiology and Immunology, 9, 61-67 null
Laboratory assessment of the mobility of water-dispersed engineered nanoparticles in a red soil (Ultisol)
Wang D, et al.
J. Hyg. (Lond), 519, 1677-1687 (2014)
Matteo Di Giosia et al.
Materials (Basel, Switzerland), 11(5) (2018-04-28)
The high hydrophobicity of fullerenes and the resulting formation of aggregates in aqueous solutions hamper the possibility of their exploitation in many technological applications. Noncovalent bioconjugation of fullerenes with proteins is an emerging approach for their dispersion in aqueous media.
Harnessing the synergistic and complementary properties of fullerene and transition-metal compounds for nanomaterial applications.
Lebedeva, et al.
Chemical Reviews, 115(20), 11301-11351 (2015)

Articles

Carbon nanotubes are materials that possess remarkable properties and offer extraordinary possibilities.

Find various photovoltaic and bioscience-based applications of fullerenes.

SWCNTs show promise in FETs, solar cells, and photodetectors due to their ultrafast charge transport mobility.

SWCNTs show promise in FETs, solar cells, and photodetectors due to their ultrafast charge transport mobility.

See All

Related Content

Batteries, fuel cells, and supercapacitors rely on electrochemical energy production. Understand their operation and electron/ion transport separation.

Batteries, fuel cells, and supercapacitors rely on electrochemical energy production. Understand their operation and electron/ion transport separation.

Batteries, fuel cells, and supercapacitors rely on electrochemical energy production. Understand their operation and electron/ion transport separation.

Batteries, fuel cells, and supercapacitors rely on electrochemical energy production. Understand their operation and electron/ion transport separation.

Our team of scientists has experience in all areas of research including Life Science, Material Science, Chemical Synthesis, Chromatography, Analytical and many others.

Contact Technical Service