Skip to Content
Merck
All Photos(2)

Key Documents

481513

Sigma-Aldrich

Selenic acid solution

40 wt. % in H2O, 99.95% trace metals basis

Sign Into View Organizational & Contract Pricing


About This Item

Linear Formula:
H2SeO4
CAS Number:
Molecular Weight:
144.97
MDL number:
UNSPSC Code:
12352300
PubChem Substance ID:
NACRES:
NA.23

Quality Level

Assay

99.95% trace metals basis

concentration

40 wt. % in H2O

density

1.407 g/mL at 25 °C

SMILES string

O[Se](O)(=O)=O

InChI

1S/H2O4Se/c1-5(2,3)4/h(H2,1,2,3,4)

InChI key

QYHFIVBSNOWOCQ-UHFFFAOYSA-N

General description

Selenic acid is a strong oxidising agent. It reacts with gold to form gold selenate.

Application

Selenic acid was used to prepare magnesium selenate.

Packaging

Packaged in glass bottles

Signal Word

Danger

Hazard Classifications

Acute Tox. 3 Inhalation - Acute Tox. 3 Oral - Aquatic Acute 1 - Aquatic Chronic 1 - Eye Dam. 1 - Skin Corr. 1B - STOT RE 2

Storage Class Code

6.1B - Non-combustible acute toxic Cat. 1 and 2 / very toxic hazardous materials

WGK

WGK 2

Flash Point(F)

Not applicable

Flash Point(C)

Not applicable


Certificates of Analysis (COA)

Search for Certificates of Analysis (COA) by entering the products Lot/Batch Number. Lot and Batch Numbers can be found on a product’s label following the words ‘Lot’ or ‘Batch’.

Already Own This Product?

Find documentation for the products that you have recently purchased in the Document Library.

Visit the Document Library

Ana Colmenero-Repiso et al.
Cancers, 12(11) (2020-11-26)
Neuroblastoma (NB) is one of the most common pediatric cancers and presents a poor survival rate in affected children. Current pretreatment risk assessment relies on a few known molecular parameters, like the amplification of the oncogene MYCN. However, a better
Structure of magnesium selenate enneahydrate, MgSeO4? 9H2O, from 5 to 250 K using neutron time-of-flight Laue diffraction
Fortes AD, et al.
Acta Crystallographica Section B, Structural Science, Crystal Engineering and Materials, 71(3) (2015)
Romain Mercier et al.
Cell, 152(5), 997-1007 (2013-03-05)
The peptidoglycan cell wall is a hallmark of the bacterial subkingdom. Surprisingly, many modern bacteria retain the ability to switch into a wall-free state called the L-form. L-form proliferation is remarkable in being independent of the normally essential FtsZ-based division
Daniel S Liu et al.
PloS one, 8(2), e52823-e52823 (2013-03-05)
Neurexin and neuroligin are transmembrane adhesion proteins that play an important role in organizing the neuronal synaptic cleft. Our lab previously reported a method for imaging the trans-synaptic binding of neurexin and neuroligin called BLINC (Biotin Labeling of INtercellular Contacts).
Gregory Bokinsky et al.
Journal of bacteriology, 195(14), 3173-3182 (2013-05-15)
Persistence is a phenomenon whereby a subpopulation of bacterial cells enters a transient growth-arrested state that confers antibiotic tolerance. While entrance into persistence has been linked to the activities of toxin proteins, the molecular mechanisms by which toxins induce growth

Our team of scientists has experience in all areas of research including Life Science, Material Science, Chemical Synthesis, Chromatography, Analytical and many others.

Contact Technical Service