All Photos(4)
About This Item
Linear Formula:
(C2H5)4BNa
CAS Number:
Molecular Weight:
150.05
MDL number:
UNSPSC Code:
12161600
PubChem Substance ID:
NACRES:
NA.22
Recommended Products
Assay
97%
reaction suitability
core: boron
reagent type: catalyst
mp
140-142 °C (lit.)
SMILES string
[Na+].CC[B-](CC)(CC)CC
InChI
1S/C8H20B.Na/c1-5-9(6-2,7-3)8-4;/h5-8H2,1-4H3;/q-1;+1
InChI key
SZSBMTRYJRHYNI-UHFFFAOYSA-N
Looking for similar products? Visit Product Comparison Guide
General description
Sodium tetraethylborate (NaBEt4) is an air and moisture-sensitive pyrophoric organoboron compound. It is very reactive in the presence of certain organometallic species and is used as an ethyl transfer reagent.
Application
Sodium tetraethylborate can be used as a derivatizing agent in the GC-MS analyses of fenbutatin oxide and alkyl tin compounds.
related product
Product No.
Description
Pricing
Signal Word
Danger
Hazard Statements
Precautionary Statements
Hazard Classifications
Pyr. Sol. 1 - Water-react 2
Storage Class Code
4.2 - Pyrophoric and self-heating hazardous materials
WGK
WGK 3
Flash Point(F)
Not applicable
Flash Point(C)
Not applicable
Personal Protective Equipment
dust mask type N95 (US), Eyeshields, Gloves
Choose from one of the most recent versions:
Already Own This Product?
Find documentation for the products that you have recently purchased in the Document Library.
Customers Also Viewed
A Sabah et al.
Chemosphere, 144, 2497-2506 (2015-12-02)
The aim of this study was to investigate the fate and behaviour of butyltin pollutants, including monobutyltin (MBT), dibutylin (DBT), and tributyltin (TBT), in waste stabilisation ponds (WSP). The study was conducted as part of a baseline survey and included
Joseph S Denkenberger et al.
Ecotoxicology (London, England), 29(10), 1614-1626 (2020-01-12)
Mercury (Hg) concentrations and speciation were measured in nine tributaries to Lake Ontario as part of two independent field-sampling programs. Among the study tributaries, mean total Hg (THg) concentrations ranged from 0.9 to 2.6 ng/L; mean dissolved Hg (THgD) ranged from
Andrea Santa-Rios et al.
Chemosphere, 266, 129001-129001 (2020-11-30)
The artisanal and small-scale gold mining (ASGM) sector uses the most mercury (Hg) worldwide. Despite health concerns associated with high Hg exposures in these communities, ASGM sites are often situated in resource limited and remote regions which challenge traditional human
Caitlyn Donadt et al.
Chemosphere, 262, 128059-128059 (2020-11-14)
Bioaccumulation of mercury in freshwater fish is a complex process driven by environmental and biological factors. In this study, we assessed mercury in fish from four tributaries to the Red Deer River, Alberta, Canada, which are characterized by high surface
Wenbin Cui et al.
Journal of agricultural and food chemistry, 65(44), 9569-9578 (2017-10-27)
Recent reports of elevated levels of methylmercury (MeHg) in rice revealed the possible occurrence of MeHg in infant rice cereals, leading to potential MeHg exposure through cereal consumption. Total mercury (THg) and MeHg levels in 119 infant cereal samples commonly
Our team of scientists has experience in all areas of research including Life Science, Material Science, Chemical Synthesis, Chromatography, Analytical and many others.
Contact Technical Service