Skip to Content
Merck
All Photos(1)

Key Documents

479624

Sigma-Aldrich

4-(Phenylazo)benzoic acid

98%

Synonym(s):

4-(2-Phenyldiazenyl)benzoic acid, 4-Carboxyazobenzene, Azobenzene-4-carboxylic acid, Azoic acid, p-Phenylazobenzoic acid

Sign Into View Organizational & Contract Pricing


About This Item

Linear Formula:
C6H5N=NC6H4CO2H
CAS Number:
Molecular Weight:
226.23
EC Number:
MDL number:
UNSPSC Code:
12352100
PubChem Substance ID:
NACRES:
NA.22

Quality Level

Assay

98%

mp

247-250 °C (lit.)

SMILES string

OC(=O)c1ccc(cc1)\N=N\c2ccccc2

InChI

1S/C13H10N2O2/c16-13(17)10-6-8-12(9-7-10)15-14-11-4-2-1-3-5-11/h1-9H,(H,16,17)/b15-14+

InChI key

CSPTZWQFHBVOLO-CCEZHUSRSA-N

General description

4-(Phenylazo)benzoic acid (PABA), an azobenzene derivative, is a photo-isomerisable molecule. It can be synthesized by reacting p-aminobenzoic acid with nitrosobenzene. The interaction of PABA with TiO2 and ZnO electrodes has been investigated. A photoreversible switch has been developed using dimethylamino calix[4]arene and PABA. The post functionalization of poly(hydroxyethyl acrylate) core cross-linked star polymers with PABA induces an ability to complex with α-cyclodextrin.

Application

4-(Phenylazo)benzoic acid may be used in the preparation of a novel photochromic ZrO2 precursor solution.

Pictograms

Exclamation mark

Signal Word

Warning

Hazard Statements

Precautionary Statements

Hazard Classifications

Eye Irrit. 2 - Skin Irrit. 2 - STOT SE 3

Target Organs

Respiratory system

Storage Class Code

11 - Combustible Solids

WGK

WGK 3

Flash Point(F)

Not applicable

Flash Point(C)

Not applicable


Certificates of Analysis (COA)

Search for Certificates of Analysis (COA) by entering the products Lot/Batch Number. Lot and Batch Numbers can be found on a product’s label following the words ‘Lot’ or ‘Batch’.

Already Own This Product?

Find documentation for the products that you have recently purchased in the Document Library.

Visit the Document Library

Synthesis of a new photochromic ZrO2 precursor for preparation of functional thin films.
Nishizawa K, et al.
Key Engineering Materials, 320, 175- 178 (2006)
Chao Luo et al.
Advanced materials (Deerfield Beach, Fla.), 30(23), e1706498-e1706498 (2018-04-25)
Organic compounds are desirable alternatives for sustainable lithium-ion battery electrodes. However, the electrochemical properties of state-of-the-art organic electrodes are still worse than commercial inorganic counterparts. Here, a new chemistry is reported based on the electrochemical conversion of nitro compounds to
Azobenzene-Functionalised Core Cross-Linked Star Polymers and their Host-Guest Interactions.
Tan S, et al.
Australian Journal of Chemistry, 67(1), 173-178 (2014)
New dyes for solar cells based on nanostructured semiconducting metal oxides: Synthesis and characterisation of ruthenium (II) complexes with thiol-substituted ligands.
Ohlsson J, et al.
Journal of Photochemistry and Photobiology A: Chemistry, 148(1), 41-48 (2002)
A photoresponsive wettability switch based on a dimethylamino Calix[4]arene.
Zhang X, et al.
Chemistry (Weinheim An Der Bergstrasse, Germany), 20(30), 9367-9371 (2014)

Our team of scientists has experience in all areas of research including Life Science, Material Science, Chemical Synthesis, Chromatography, Analytical and many others.

Contact Technical Service