Skip to Content
Merck
All Photos(1)

Key Documents

479330

Sigma-Aldrich

Lithium chloride/potassium chloride eutectic

AnhydroBeads, 99.99% trace metals basis

Synonym(s):

Lithium potassium chloride

Sign Into View Organizational & Contract Pricing


About This Item

CAS Number:
MDL number:
UNSPSC Code:
12352302
PubChem Substance ID:
NACRES:
NA.23

product line

AnhydroBeads

Quality Level

Assay

99.99% trace metals basis

composition

LiCl, 44 wt. %

impurities

≤150.0 ppm Trace Metal Analysis

mp

357 °C (lit.)

density

2.02 g/mL at 25 °C (lit.)

SMILES string

[Li+].[Cl-].[Cl-].[K+]

InChI

1S/2ClH.K.Li/h2*1H;;/q;;2*+1/p-2

InChI key

HCQWRNRRURULEY-UHFFFAOYSA-L

Looking for similar products? Visit Product Comparison Guide

General description

Lithium chloride/potassium chloride eutectic(LKE) can serve as a medium for chemical and electrochemical processing. It has potential application in the field of molten salt batteries, metal refining, pyrochemical nuclear fuel processing and electric power cells.

Application

LKE can be used to prepare:
  • Microporous separator pellets for primary Li thermal batteries.
  • Electrolytic bath for the separation of actinides from lanthanides on aluminum electrode.
  • Electrolyte for the reduction and electrorefining of Uranium.

Legal Information

AnhydroBeads is a trademark of Sigma-Aldrich Co. LLC

Pictograms

Exclamation mark

Signal Word

Warning

Hazard Statements

Hazard Classifications

Acute Tox. 4 Oral - Eye Irrit. 2 - Skin Irrit. 2

Storage Class Code

13 - Non Combustible Solids

WGK

WGK 1

Flash Point(F)

Not applicable

Flash Point(C)

Not applicable

Personal Protective Equipment

dust mask type N95 (US), Eyeshields, Gloves

Choose from one of the most recent versions:

Certificates of Analysis (COA)

Lot/Batch Number

Don't see the Right Version?

If you require a particular version, you can look up a specific certificate by the Lot or Batch number.

Already Own This Product?

Find documentation for the products that you have recently purchased in the Document Library.

Visit the Document Library

Customers Also Viewed

Slide 1 of 1

1 of 1

Guidotti, R. Reinhardt, F.W.
Proc. 37th Power Sources Conf., 251-251 (1996)

Our team of scientists has experience in all areas of research including Life Science, Material Science, Chemical Synthesis, Chromatography, Analytical and many others.

Contact Technical Service