Skip to Content
Merck
All Photos(2)

Key Documents

472956

Sigma-Aldrich

Tungsten hexacarbonyl

99.99% trace metals basis (excluding Mo), purified by sublimation

Synonym(s):

Hexacarbonyltungsten, Tungsten carbonyl, Tungsten hexacarbonyl

Sign Into View Organizational & Contract Pricing


About This Item

Linear Formula:
W(CO)6
CAS Number:
Molecular Weight:
351.90
EC Number:
MDL number:
UNSPSC Code:
12352103
PubChem Substance ID:
NACRES:
NA.23

vapor density

12.1 (vs air)

Quality Level

vapor pressure

1.2 mmHg ( 67 °C)

Assay

99.99% trace metals basis (excluding Mo)

form

solid

purified by

sublimation

reaction suitability

core: tungsten
reagent type: catalyst

mp

150 °C (lit.)

density

2.65 g/mL at 25 °C (lit.)

SMILES string

[W].[C-]#[O+].[C-]#[O+].[C-]#[O+].[C-]#[O+].[C-]#[O+].[C-]#[O+]

InChI

1S/6CO.W/c6*1-2;

InChI key

FQNHWXHRAUXLFU-UHFFFAOYSA-N

Looking for similar products? Visit Product Comparison Guide

General description

Atomic number of base material: 74 Tungsten

Application

A precursor for the perparation of tungsten-containing complexes or nanomaterials. Also used in photochemistry
Catalyst used in the photochemical cyclization of silyloxytrienes via either a 6-endo pathway or a Cope rearrangement.

Pictograms

Skull and crossbones

Signal Word

Danger

Hazard Statements

Hazard Classifications

Acute Tox. 3 Dermal - Acute Tox. 3 Inhalation - Acute Tox. 3 Oral

Storage Class Code

6.1D - Non-combustible acute toxic Cat.3 / toxic hazardous materials or hazardous materials causing chronic effects

WGK

WGK 3

Flash Point(F)

Not applicable

Flash Point(C)

Not applicable

Personal Protective Equipment

dust mask type N95 (US), Eyeshields, Gloves

Choose from one of the most recent versions:

Certificates of Analysis (COA)

Lot/Batch Number

Don't see the Right Version?

If you require a particular version, you can look up a specific certificate by the Lot or Batch number.

Already Own This Product?

Find documentation for the products that you have recently purchased in the Document Library.

Visit the Document Library

Denny, N.R.
Proc. SPIE: Int. Soc. Opt. Eng., 6005-6005 (2005)
Journal of Organometallic Chemistry, 692, 562-562 (2007)
Utke, I.; et al.
Advanced Engineering Materials, 8 (3), 155-157 (2006)
Kuramshin, A.I. et al.
Russ. J. Org. Chem., 41(5), 649-655 (2005)
Seth M Dumbris et al.
The Journal of organic chemistry, 74(22), 8862-8865 (2009-10-30)
Hydantoins can be synthesized from the corresponding amino amides employing oxidative catalytic carbonylation using W(CO)(6) as the catalyst, I(2) as the oxidant, CO as the carbonyl source, and DBU as base. Secondary amides afford the hydantoins in good to excellent

Articles

Thin film photovoltaic devices have become increasingly important in efficiently harnessing solar energy to meet consumer demand.

The properties of many devices are limited by the intrinsic properties of the materials that compose them.

Our team of scientists has experience in all areas of research including Life Science, Material Science, Chemical Synthesis, Chromatography, Analytical and many others.

Contact Technical Service