Skip to Content
Merck
All Photos(1)

Key Documents

469114

Sigma-Aldrich

6-Nitrobenzothiazole

99%

Sign Into View Organizational & Contract Pricing


About This Item

Empirical Formula (Hill Notation):
C7H4N2O2S
CAS Number:
Molecular Weight:
180.18
EC Number:
MDL number:
UNSPSC Code:
12352100
PubChem Substance ID:
NACRES:
NA.22

Quality Level

Assay

99%

mp

175-178 °C (lit.)

SMILES string

[O-][N+](=O)c1ccc2ncsc2c1

InChI

1S/C7H4N2O2S/c10-9(11)5-1-2-6-7(3-5)12-4-8-6/h1-4H

InChI key

QLUFBCVWKTWKBF-UHFFFAOYSA-N

General description

The XCORFE (Xnucleus-proton correlation with fixed evolution time) pulse sequence of 6-nitrobenzothiazole has been tested.

Application

6-Nitrobenzothiazole may be used in the preparation of 4-amino-3-sulfanylbenzonitrile and 2-amino-5-nitrobenzothiole, via alkaline hydrolysis.

Pictograms

Exclamation mark

Signal Word

Warning

Hazard Statements

Hazard Classifications

Eye Irrit. 2 - Skin Irrit. 2 - STOT SE 3

Target Organs

Respiratory system

Storage Class Code

11 - Combustible Solids

WGK

WGK 3

Flash Point(F)

Not applicable

Flash Point(C)

Not applicable

Personal Protective Equipment

dust mask type N95 (US), Eyeshields, Gloves

Certificates of Analysis (COA)

Search for Certificates of Analysis (COA) by entering the products Lot/Batch Number. Lot and Batch Numbers can be found on a product’s label following the words ‘Lot’ or ‘Batch’.

Already Own This Product?

Find documentation for the products that you have recently purchased in the Document Library.

Visit the Document Library

Heterocyclic Amplifiers of Phleomycin. IX. Some Derivatives of Fused and Unfused Mono-and Di-aza Heterocycles.
Barlin GB and Ireland SJ.
Australian Journal of Chemistry, 38(11), 1685-1691 (1985)
Assignment of quaternary carbons in aromatic compounds by long-range heteronuclear shift correlated 2D-NMR spectroscopy and its application to acteoside.
Numata A, et al.
Agricultural and Biological Chemistry, 51(4), 1199-1201 (1987)
Livio Racané et al.
Bioorganic & medicinal chemistry, 18(3), 1038-1044 (2010-01-12)
The efficient synthesis of new bis-substituted nitro-amidino, amino-amidino (10a, 10b-13a, 13b) and previously prepared diamidino 2-phenyl-benzothiazoles (9a, 9b) is described. The compounds 11a and 11b were prepared by recently developed methodology of the key precursors in zwitterionic form 8a and

Our team of scientists has experience in all areas of research including Life Science, Material Science, Chemical Synthesis, Chromatography, Analytical and many others.

Contact Technical Service