468134
Poly(styrene-co-allyl alcohol)
average Mw ~2,200, average Mn ~1,200, allyl alcohol 40 mol %
Synonym(s):
Allyl alcohol-styrene copolymer, Poly(styrene-allyl alcohol), Styrene-allyl alcohol copolymer
About This Item
Recommended Products
vapor density
2 (vs air)
Quality Level
vapor pressure
23.8 mmHg ( 25 °C)
form
solid
mol wt
average Mn ~1,200
average Mw ~2,200
composition
allyl alcohol, 40 mol %
expl. lim.
18 %
hydroxyl value
255 mg KOH/g
transition temp
Tg 63 °C (DSC)
density
1.05 g/mL at 25 °C
SMILES string
OCC=C.C=Cc1ccccc1
InChI
1S/C8H8.C3H6O/c1-2-8-6-4-3-5-7-8;1-2-3-4/h2-7H,1H2;2,4H,1,3H2
InChI key
WXNYILVTTOXAFR-UHFFFAOYSA-N
Looking for similar products? Visit Product Comparison Guide
Related Categories
Application
- Reinforced Shape-tunable Microwrinkles: Uses Poly(styrene-co-allyl alcohol) as a sacrificial coating to create reinforced, shape-tunable microwrinkles on elastomer surfaces, enhancing surface functionality for various applications (T Ohzono et al., researchgate.net).
- Electronic Nose for VOC Detection: Describes the use of Poly(styrene-co-allyl alcohol) in sensor arrays for selective detection of volatile organic compounds (VOCs) via principal component analysis (I Yoon et al., Sensors, 2019).
- Proton Conductive Membranes: Focuses on developing proton conductive membranes from Poly(styrene-co-allyl alcohol) for potential use in fuel cells, highlighting the impact of hydroxyl group interactions on conductivity (FAM Loureiro et al., Polímeros, 2014).
- Gas Separation Membranes: Utilizes Poly(styrene-co-allyl alcohol) in blend membranes with polycarbonate, enhanced with nano-zeolite for improved gas separation efficiency (A Kausar, Journal of the Chinese Advanced Materials Society, 2017).
Storage Class Code
11 - Combustible Solids
WGK
WGK 3
Flash Point(F)
Not applicable
Flash Point(C)
Not applicable
Personal Protective Equipment
Choose from one of the most recent versions:
Already Own This Product?
Find documentation for the products that you have recently purchased in the Document Library.
Customers Also Viewed
Our team of scientists has experience in all areas of research including Life Science, Material Science, Chemical Synthesis, Chromatography, Analytical and many others.
Contact Technical Service