Skip to Content
Merck
All Photos(3)

Key Documents

468096

Sigma-Aldrich

Poly(tetrafluoroethylene)

powder, 35 μm particle size

Synonym(s):

Fluorinated polymer, Fluoropolymer, PTFE

Sign Into View Organizational & Contract Pricing


About This Item

Linear Formula:
(CF2CF2)n
CAS Number:
MDL number:
UNSPSC Code:
12162002
PubChem Substance ID:
NACRES:
NA.23
Pricing and availability is not currently available.

form

powder

Quality Level

particle size

35 μm

transition temp

Tm 321 °C

density

2.1 g/mL at 25 °C

application(s)

battery manufacturing

SMILES string

F\C(F)=C(\F)F

InChI

1S/C2F4/c3-1(4)2(5)6

InChI key

BFKJFAAPBSQJPD-UHFFFAOYSA-N

Looking for similar products? Visit Product Comparison Guide

Storage Class Code

11 - Combustible Solids

WGK

WGK 3

Flash Point(F)

Not applicable

Flash Point(C)

Not applicable

Personal Protective Equipment

dust mask type N95 (US), Eyeshields, Gloves

Choose from one of the most recent versions:

Certificates of Analysis (COA)

Lot/Batch Number

Don't see the Right Version?

If you require a particular version, you can look up a specific certificate by the Lot or Batch number.

Already Own This Product?

Find documentation for the products that you have recently purchased in the Document Library.

Visit the Document Library

Jianli Zou et al.
Nature communications, 5, 5254-5254 (2014-10-17)
Despite recent progress in preparing numerous types of nanosheets, it remains a difficult challenge to assemble the tiny building blocks into functional macroscale architectures suitable for practical applications. Here we introduce a diffusion driven layer-by-layer assembly process and demonstrate its
Tobias Fink et al.
Anesthesiology, 122(1), 117-126 (2014-08-30)
Multicapillary column ion-mobility spectrometry (MCC-IMS) may identify volatile components in exhaled gas. The authors therefore used MCC-IMS to evaluate exhaled gas in a rat model of sepsis, inflammation, and hemorrhagic shock. Male Sprague-Dawley rats were anesthetized and ventilated via tracheostomy
Hocine Bensaadi et al.
The American surgeon, 80(1), 57-65 (2014-01-10)
Funding received from Cousin Biotech, Wervicq Sud, France, and CR Bard Inc., Cranston, RI. The aim of this prospective randomized study was to determine the long-term recurrence and complication rates after small abdominal wall hernia repair with two different bilayer
Ming Yang et al.
The Journal of thoracic and cardiovascular surgery, 150(1), 152-157 (2015-03-15)
Robotically assisted cardiac surgery is an alternative to conventional, open-chest surgery. Although studies have been done on the clinical effect, morbidity, and mortality of robotically assisted atrial myxoma excision, few have addressed surgical outcomes, such as pain, quality of life
René R E Steendam et al.
Nature communications, 5, 5543-5543 (2014-11-22)
The synthesis of enantiopure molecules from achiral precursors without the need for pre-existing chirality is a major challenge associated with the origin of life. We here show that an enantiopure product can be obtained from achiral starting materials in a

Articles

Polymer electrolyte membrane (PEM) fuel cells offer high efficiency and low-temperature operation, reducing oil use in light-duty vehicle transportation.

Polymer electrolyte membrane (PEM) fuel cells offer high efficiency and low-temperature operation, reducing oil use in light-duty vehicle transportation.

Polymer electrolyte membrane (PEM) fuel cells offer high efficiency and low-temperature operation, reducing oil use in light-duty vehicle transportation.

Polymer electrolyte membrane (PEM) fuel cells offer high efficiency and low-temperature operation, reducing oil use in light-duty vehicle transportation.

Questions

  1. Mean particle diameter가 35 μm라고 specification sheet에 적혀있는데, 이 기준이 정확히 무엇인지 궁금합니다. 각 particle이 직경 35 μm인 구와 동일한 부피를 가진 것일까요? 아니면, 2D 평면에서의 단면적이 직경 35 μm인 원과 동일한 것일까요? 감사합니다.

    1 answer
    1. The size is determined by a dynamic light scattering technique. This method measures fluctuations in the intensity of scattered light from a sample, which are directly related to the Brownian motion of the particles. Smaller particles move faster, causing more rapid fluctuations in the scattered light. This allows for the calculation of their diffusion coefficient and subsequently their size using the Stokes-Einstein equation.

      Helpful?

Reviews

No rating value

Active Filters

Our team of scientists has experience in all areas of research including Life Science, Material Science, Chemical Synthesis, Chromatography, Analytical and many others.

Contact Technical Service