Skip to Content
Merck
All Photos(2)

Key Documents

466948

Sigma-Aldrich

1-(p-Tolylsulfonyl)pyrrole

97%

Sign Into View Organizational & Contract Pricing


About This Item

Empirical Formula (Hill Notation):
C11H11NO2S
CAS Number:
Molecular Weight:
221.28
MDL number:
UNSPSC Code:
12352100
PubChem Substance ID:
NACRES:
NA.22

Assay

97%

form

solid

mp

99-102 °C (lit.)

SMILES string

Cc1ccc(cc1)S(=O)(=O)n2cccc2

InChI

1S/C11H11NO2S/c1-10-4-6-11(7-5-10)15(13,14)12-8-2-3-9-12/h2-9H,1H3

InChI key

OXWIEWFMRVJGNY-UHFFFAOYSA-N

General description

1-(p-Tolylsulfonyl)pyrrole is a pyrrole derivative. Its Fourier-Transform-Infrared, Fourier-Transform-Raman and NMR spectra have been studied.

Application

1-(p-Tolylsulfonyl)pyrrole may be used in the synthesis of 4-(3-pyrrolyl) butanoic aicd (PBA).

Pictograms

Exclamation mark

Signal Word

Warning

Hazard Statements

Hazard Classifications

Eye Irrit. 2 - Skin Irrit. 2 - STOT SE 3

Target Organs

Respiratory system

Storage Class Code

11 - Combustible Solids

WGK

WGK 3

Flash Point(F)

Not applicable

Flash Point(C)

Not applicable

Personal Protective Equipment

dust mask type N95 (US), Eyeshields, Gloves

Choose from one of the most recent versions:

Certificates of Analysis (COA)

Lot/Batch Number

Don't see the Right Version?

If you require a particular version, you can look up a specific certificate by the Lot or Batch number.

Already Own This Product?

Find documentation for the products that you have recently purchased in the Document Library.

Visit the Document Library

Y Erdogdu et al.
Spectrochimica acta. Part A, Molecular and biomolecular spectroscopy, 146, 88-96 (2015-03-31)
Fourier-Transform-Infrared, Fourier-Transform-Raman and Nuclear Magnetic Rezonans spectra of 1-(p-tolylsulfonyl) pyrrole molecule have been recorded. In the powder form, vibrational spectra of 1-(p-tolylsulfonyl) pyrrol molecule were investigated in the region 4000-400 cm(-1) and 3500-50 cm(-1), respectively. The conformational analysis, geometrical structure
Hui Peng et al.
Biosensors & bioelectronics, 20(9), 1821-1828 (2005-02-01)
A simple and label-free electrochemical sensor for recognition of the DNA hybridization event was prepared based on a new functionalised conducting copolymer, poly[pyrrole-co-4-(3-pyrrolyl) butanoic acid]. This precursor copolymer can be easily electrodeposited on the electrode surface and shows high electroactivity

Our team of scientists has experience in all areas of research including Life Science, Material Science, Chemical Synthesis, Chromatography, Analytical and many others.

Contact Technical Service