Skip to Content
Merck
All Photos(2)

Key Documents

466867

Sigma-Aldrich

Polybutadiene, predominantly 1,2-addition

approx. 90% 1,2-vinyl

Sign Into View Organizational & Contract Pricing


About This Item

Linear Formula:
[CH2CH(CH=CH2)]n
CAS Number:
MDL number:
UNSPSC Code:
12162002
PubChem Substance ID:
NACRES:
NA.23

Quality Level

refractive index

n20/D 1.5002 (lit.)

viscosity

30-100 poise(45 °C)(lit.)

transition temp

Tg −30 °C

density

0.86 g/mL at 25 °C (lit.)

SMILES string

C=CC=C

InChI

1S/C4H8/c1-3-4-2/h3-4H,1-2H3/b4-3+

InChI key

IAQRGUVFOMOMEM-ONEGZZNKSA-N

Application

  • Transforming Polybutadiene with Tetrazine Click Chemistry into Antioxidant Foams That Fluoresce with Oxidation: This research uses tetrazine click chemistry to modify polybutadiene, enhancing its properties to create antioxidant foams that respond to oxidation with fluorescence, demonstrating innovative ways to utilize 1,2-addition polybutadiene (RE Bagge et al., 2017).
  • Isoxazoline-based porous polymer for the highly effective adsorption of 2,4,6-trinitrotoluene (TNT): Catalyst-free click polymerization between an in situ generated nitrile oxide with polybutadiene. This article explores the enhancement of polybutadiene′s adsorptive properties for environmental cleanup applications, emphasizing how its structural characteristics affect performance (H Deng et al., 2020).
  • Modification of polybutadiene with trifluoromethyl and clickable azide groups in one shot: This paper describes the simultaneous introduction of trifluoromethyl and azide functional groups into polybutadiene, showcasing the versatility of 1,2-addition polybutadiene in chemical modifications (S Wang et al., 2021).

Storage Class Code

10 - Combustible liquids

WGK

WGK 3

Flash Point(F)

Not applicable

Flash Point(C)

Not applicable

Personal Protective Equipment

dust mask type N95 (US), Eyeshields, Gloves

Certificates of Analysis (COA)

Search for Certificates of Analysis (COA) by entering the products Lot/Batch Number. Lot and Batch Numbers can be found on a product’s label following the words ‘Lot’ or ‘Batch’.

Already Own This Product?

Find documentation for the products that you have recently purchased in the Document Library.

Visit the Document Library

Zhi-ping Huang et al.
Journal of hazardous materials, 229-230, 251-257 (2012-06-26)
Migration appeared in the interfaces of nitrate ester plasticized polyether (NEPE) based propellant/hydroxyl-terminated polybutadiene (HTPB) based liner/ethylene propylene terpolymer (EPDM) based insulation was studied by aging at different temperatures. The migration components were extracted with solvent and determined by high
Li Li et al.
ACS nano, 5(10), 7754-7766 (2011-08-27)
Understanding the relevant permeability properties of ultrafiltration membranes is facilitated by using materials and procedures that allow a high degree of control on morphology and chemical composition. Here we present the first study on diffusion permeability through gyroid nanoporous cross-linked
Rachel Calvet et al.
Journal of chromatography. A, 1253, 164-170 (2012-07-24)
In this study, the coupling of IGC-ID analysis and carbon blacks impregnation with increasing ratios of polymer allow us to highlight the affinity of the polymer to the solid surface. The variations of the dispersive component of the surface energy
P S Brown et al.
Langmuir : the ACS journal of surfaces and colloids, 28(38), 13712-13719 (2012-09-13)
Two-dimensional hexagonally ordered honeycomb surfaces have been created by solvent casting polybutadiene films under controlled humidity. Subsequent CF(4) plasmachemical fluorination introduces cross-linking and surface texturing, leading to hierarchical surfaces with roughness on both the 10 μm (honeycomb) and micrometer (texturing)
Gregory P Robbins et al.
Langmuir : the ACS journal of surfaces and colloids, 26(17), 14089-14096 (2010-08-14)
The polymersome, a fully synthetic cell mimetic, is a tunable platform for drug delivery vehicles to detect and treat disease (theranostics). Here, we design a leuko-polymersome, a polymersome with the adhesive properties of leukocytes, which can effectively bind to inflammatory

Our team of scientists has experience in all areas of research including Life Science, Material Science, Chemical Synthesis, Chromatography, Analytical and many others.

Contact Technical Service