Skip to Content
Merck
All Photos(1)

Key Documents

466069

Sigma-Aldrich

Ytterbium

powder, ≥99.9% trace rare earth metals basis

Sign Into View Organizational & Contract Pricing


About This Item

Empirical Formula (Hill Notation):
Yb
CAS Number:
Molecular Weight:
173.04
EC Number:
MDL number:
UNSPSC Code:
12161600
PubChem Substance ID:
NACRES:
NA.23

Quality Level

Assay

≥99.9% trace rare earth metals basis

form

powder

reaction suitability

reagent type: catalyst
core: ytterbium

resistivity

28 μΩ-cm, 20°C

bp

1196 °C (lit.)

mp

824 °C (lit.)

density

6.54 g/mL at 25 °C (lit.)

SMILES string

[Yb]

InChI

1S/Yb

InChI key

NAWDYIZEMPQZHO-UHFFFAOYSA-N

Looking for similar products? Visit Product Comparison Guide

Application

When reacted with aromatic aldimines, followed by oxidants, produces vicinal diimine compounds.

Pictograms

FlameExclamation mark

Signal Word

Danger

Hazard Classifications

Acute Tox. 4 Dermal - Acute Tox. 4 Inhalation - Acute Tox. 4 Oral - Eye Irrit. 2 - Flam. Sol. 1 - Skin Irrit. 2 - STOT SE 3

Target Organs

Respiratory system

Storage Class Code

4.1B - Flammable solid hazardous materials

WGK

WGK 3

Flash Point(F)

Not applicable

Flash Point(C)

Not applicable

Personal Protective Equipment

dust mask type N95 (US), Eyeshields, Gloves

Choose from one of the most recent versions:

Certificates of Analysis (COA)

Lot/Batch Number

Don't see the Right Version?

If you require a particular version, you can look up a specific certificate by the Lot or Batch number.

Already Own This Product?

Find documentation for the products that you have recently purchased in the Document Library.

Visit the Document Library

Customers Also Viewed

Jin, W-S. et al.
Chemical Communications (Cambridge, England), 1101-1101 (1998)
Jianhua Wang et al.
Optics express, 20(27), 28373-28378 (2012-12-25)
A 100 W-class all-fiber linearly-polarized single-mode fiber laser at 1120 nm with an optical efficiency of 50% was demonstrated. The laser consists of a 4.2 m long Yb-doped polarization maintaining fiber with a core diameter of 10 μm and a
Tao Zhang et al.
Chemistry (Weinheim an der Bergstrasse, Germany), 19(2), 739-748 (2012-11-21)
Based on a donor-acceptor framework, several conjugates have been designed and prepared in which an electron-donor moiety, ytterbium(III) porphyrinate (YbPor), was linked through an ethynyl bridge to an electron-acceptor moiety, boron dipyrromethene (BODIPY). Photoluminescence studies demonstrated efficient energy transfer from
S Rydberg et al.
Optics express, 21(6), 6681-6688 (2013-04-03)
In this work we present experimental evidence that the valence instability of the ytterbium ion play a key role for the observed photodarkening mechanism in Yb-doped fiber lasers. Luminescence and excitation spectroscopy performed on UV irradiated Yb/Al doped silica glass
Marcus L Cole et al.
Chemistry (Weinheim an der Bergstrasse, Germany), 19(4), 1410-1420 (2012-12-04)
New reactive, divalent lanthanoid formamidinates [Yb(Form)(2)(thf)(2)] (Form=[RNCHNR]; R=o-MeC(6)H(4) (o-TolForm; 1), 2,6-Me(2)C(6)H(3) (XylForm; 2), 2,4,6-Me(3)C(6)H(2) (MesForm; 3), 2,6-Et(2)C(6)H(3) (EtForm; 4), o-PhC(6)H(4) (o-PhPhForm; 5), 2,6-iPr(2)C(6)H(3) (DippForm; 6), o-HC(6)F(4) (TFForm; 7)) and [Eu(DippForm)(2)(thf)(2)] (8) have been prepared by redox transmetallation/protolysis reactions between an

Articles

A significant limiting factor for wearable electronics and wireless sensors is the finite amount of energy that can be stored in on-board batteries.

Permanent magnets are an essential technology for energy conversion. Motors and generators are used to convert energy between electrical and mechanical forms.

Rechargeable solid-state batteries are becoming increasingly important due to wide-spread use in computers, portable electronics, and vehicular applications.

Rechargeable solid-state batteries are becoming increasingly important due to wide-spread use in computers, portable electronics, and vehicular applications.

See All

Our team of scientists has experience in all areas of research including Life Science, Material Science, Chemical Synthesis, Chromatography, Analytical and many others.

Contact Technical Service