Skip to Content
Merck
All Photos(1)

Key Documents

460400

Sigma-Aldrich

Titanium

wire, diam. 0.25 mm, 99.7% trace metals basis

Sign Into View Organizational & Contract Pricing


About This Item

Empirical Formula (Hill Notation):
Ti
CAS Number:
Molecular Weight:
47.87
EC Number:
MDL number:
UNSPSC Code:
12141746
PubChem Substance ID:
NACRES:
NA.23
Pricing and availability is not currently available.

Quality Level

Assay

99.7% trace metals basis

form

wire

autoignition temp.

860 °F

resistivity

42.0 μΩ-cm, 20°C

diam.

0.25 mm

bp

3287 °C (lit.)

mp

1660 °C (lit.)

density

4.5 g/mL at 25 °C (lit.)

SMILES string

[Ti]

InChI

1S/Ti

InChI key

RTAQQCXQSZGOHL-UHFFFAOYSA-N

General description

Titanium (Ti) wire is a semiconductor material that has an energy band gap of ~4.85 eV and an electrical conductivity of ~10-13 Ω-1cm-1. [1]

Application

Ti wire can be used in a variety of applications such as dye sensitized solar cells (DSSCs),[2] solid phase microextraction fiber,[3] biomedical use,[4] and in the fabrication of TiO2 nanotubes.[5]

Quantity

2.2 g = 10 m; 11 g = 50 m

Storage Class Code

11 - Combustible Solids

WGK

nwg

Flash Point(F)

Not applicable

Flash Point(C)

Not applicable

Personal Protective Equipment

dust mask type N95 (US), Eyeshields, Gloves

Choose from one of the most recent versions:

Certificates of Analysis (COA)

Lot/Batch Number

Don't see the Right Version?

If you require a particular version, you can look up a specific certificate by the Lot or Batch number.

Already Own This Product?

Find documentation for the products that you have recently purchased in the Document Library.

Visit the Document Library

Improved photocatalytic degradation of textile dye using titanium dioxide nanotubes formed over titanium wires
Kar A, et al.
Environmental Science & Technology, 43(9), 3260-3265 (2009)
Porous titanium materials with entangled wire structure for load-bearing biomedical applications
He G, et al.
Journal of the Mechanical Behavior of Biomedical Materials, 5(1), 16-31 (2012)
Unbreakable solid-phase microextraction fibers obtained by sol- gel deposition on titanium wire
Azenha MA, et al.
Analytical Chemistry, 78(6), 2071-2074 (2006)
The 3-dimensional dye-sensitized solar cell and module based on all titanium substrates
Liu Y, et al.
Applied Energy, 87(2), 436-441 (2010)
Electrode coatings from sprayed titanium dioxide nanoparticles-behaviour in NaOH solutions
Hayden BE, et al.
Electrochemical Communications, 3(8), 390-394 (2001)

Articles

Biomedical implants are essentially foreign substances within the human body that must survive many years’ exposure to demanding mechanical and physiological conditions. Despite these challenges, metal implants have been widely used to substitute for or rebuild hard tissues such as bones and teeth.

Questions

Reviews

No rating value

Active Filters

Our team of scientists has experience in all areas of research including Life Science, Material Science, Chemical Synthesis, Chromatography, Analytical and many others.

Contact Technical Service