Skip to Content
Merck
All Photos(1)

Key Documents

460214

Sigma-Aldrich

Diisodecyl adipate

technical grade

Synonym(s):

DIDA (plasticizer)

Sign Into View Organizational & Contract Pricing


About This Item

Linear Formula:
C10H21O2C(CH2)4CO2C10H21
CAS Number:
Molecular Weight:
426.67
EC Number:
MDL number:
UNSPSC Code:
12162002
PubChem Substance ID:
NACRES:
NA.23

grade

technical grade

Quality Level

form

liquid

refractive index

n20/D 1.452 (lit.)

mp

27.4 °C (lit.)

solubility

hexane, toluene, ethanol and acetone: soluble
water and propylene glycol: insoluble

density

0.916 g/mL at 25 °C (lit.)

SMILES string

CC(C)CCCCCCCOC(=O)CCCCC(=O)OCCCCCCCC(C)C

InChI

1S/C26H50O4/c1-23(2)17-11-7-5-9-15-21-29-25(27)19-13-14-20-26(28)30-22-16-10-6-8-12-18-24(3)4/h23-24H,5-22H2,1-4H3

InChI key

YKGYQYOQRGPFTO-UHFFFAOYSA-N

Looking for similar products? Visit Product Comparison Guide

Application

Diisodecyl adipate is an adipate-based plasticizer that can be used in the development of biodegradable films using thermoplastic starch and poly(lactic acid). It may be used in the preparation of cellulose triacetate based films for the fabrication of optical devices. Diisodecyl adipate may form a blend with PVC to form polymeric membranes on electrodes for electrochemical applications.

Storage Class Code

10 - Combustible liquids

WGK

awg

Flash Point(F)

429.8 °F - closed cup

Flash Point(C)

221 °C - closed cup

Personal Protective Equipment

dust mask type N95 (US), Eyeshields, Gloves

Choose from one of the most recent versions:

Certificates of Analysis (COA)

Lot/Batch Number

Don't see the Right Version?

If you require a particular version, you can look up a specific certificate by the Lot or Batch number.

Already Own This Product?

Find documentation for the products that you have recently purchased in the Document Library.

Visit the Document Library

Development of microporous structure and its application to optical film for cellulose triacetate containing diisodecyl adipate
Shimada H, et al.
Carbohydrate Polymers, 120(1), 22-28 (2015)
Silver (I)-selective membrane electrodes based onmono-to quadridentate podands
Lee SS, et al.
Analyst, 123(2), 383-386 (1998)
M A Shirai et al.
Carbohydrate polymers, 92(1), 19-22 (2012-12-12)
Biodegradable films were produced from blends contained a high amount of thermoplastic starch (TPS) and poly(lactic acid) (PLA) plasticized with different adipate or citrate esters. It was not possible to obtain pellets for the production of films using only glycerol

Our team of scientists has experience in all areas of research including Life Science, Material Science, Chemical Synthesis, Chromatography, Analytical and many others.

Contact Technical Service