Skip to Content
Merck
All Photos(1)

Key Documents

455849

Sigma-Aldrich

Ammonium hydrogensulfate

99.99% trace metals basis

Synonym(s):

Ammonium bisulfate, Ammonium sulfate monobasic

Sign Into View Organizational & Contract Pricing


About This Item

Linear Formula:
(NH4)HSO4
CAS Number:
Molecular Weight:
115.11
EC Number:
MDL number:
UNSPSC Code:
12352302
PubChem Substance ID:
NACRES:
NA.23

Quality Level

Assay

99.99% trace metals basis

form

crystalline

impurities

≤200 mg/kg Trace metallic impurities analysis (ICP)

bp

350 °C (dec.)(lit.)

mp

121-145 °C (lit.)

density

1.79 g/mL at 25 °C (lit.)

SMILES string

N.OS(O)(=O)=O

InChI

1S/H3N.H2O4S/c;1-5(2,3)4/h1H3;(H2,1,2,3,4)

InChI key

BIGPRXCJEDHCLP-UHFFFAOYSA-N

Looking for similar products? Visit Product Comparison Guide

Application

  • Enhancement of Aesthetic Dental CAD-CAM Materials through Surface Etching with a Mixed Aqueous Solution of Ammonium Fluoride and Ammonium Hydrogen Sulfate - This study explores the potential of ammonium hydrogen sulfate in surface etching applications for dental materials, focusing on its low toxicity and effective etching capabilities (Y Nishizawa et al., 2024).
  • Thermodynamics of ammonioalunite precipitation in ammonium aluminum sulfate solution - Investigates the thermodynamic properties of ammonium aluminum sulfate solutions, providing insights into chemical processes involving ammonium hydrogen sulfate (X Yang et al., 2020).
  • Hygroscopic behavior and chemical composition evolution of internally mixed aerosols composed of oxalic acid and ammonium sulfate - Studies the hygroscopic properties of mixed aerosol particles, including those formed with ammonium hydrogen sulfate, to understand atmospheric chemical processes better (X Wang et al., 2017).

Pictograms

Corrosion

Signal Word

Danger

Hazard Statements

Hazard Classifications

Eye Dam. 1 - Skin Corr. 1B

Storage Class Code

8B - Non-combustible corrosive hazardous materials

WGK

WGK 1

Flash Point(F)

Not applicable

Flash Point(C)

Not applicable

Personal Protective Equipment

dust mask type N95 (US), Eyeshields, Gloves

Choose from one of the most recent versions:

Certificates of Analysis (COA)

Lot/Batch Number

Don't see the Right Version?

If you require a particular version, you can look up a specific certificate by the Lot or Batch number.

Already Own This Product?

Find documentation for the products that you have recently purchased in the Document Library.

Visit the Document Library

Sangil Lee et al.
Journal of the Air & Waste Management Association (1995), 57(9), 1123-1135 (2007-10-05)
Particulate matter (PM) less than 2.5 microm in size (PM2.5) source apportionment by chemical mass balance receptor modeling was performed to enhance regional characterization of source impacts in the southeastern United States. Secondary particles, such as NH4HSO4, (NH4)2SO4, NH4NO3, and
R B Schlesinger
Environmental health perspectives, 79, 121-126 (1989-02-01)
The ability of the lungs to clear deposited material is essential for maintenance of lung homeostasis. Acid aerosols have been shown to alter the efficiency of this process. This paper assesses the role of acid aerosol exposure concentration (C), exposure
W J Mautz et al.
Toxicological sciences : an official journal of the Society of Toxicology, 61(2), 331-341 (2001-05-16)
The purpose of this study was to examine a broad range of toxicologic responses in rats exposed to a multi-component pollutant atmosphere. Cumulative and adaptive respiratory tract responses to 3 concentrations of an inhaled particle-oxidant mixture were examined in Fisher
W S Tunnicliffe et al.
Occupational and environmental medicine, 60(11), e15-e15 (2003-10-24)
To explore the effects in normal and asthmatic adults of exposure to 200 ppb sulphur dioxide (SO2) and 200 microg/m3 and 2000 microg/m3 aerosols of ammonium bisulphate (AB) and sulphuric acid (SA) (MMD 0.3 microm). Exposures were placebo controlled, for
Bryan R Bzdek et al.
The journal of physical chemistry. A, 114(43), 11638-11644 (2010-10-13)
The reaction kinetics of ammonium bisulfate clusters with dimethylamine (DMA) gas were investigated using Fourier transform ion cyclotron resonance mass spectrometry (FTICR-MS). Clusters ranged in size from 1 to 10 bisulfate ions. Although displacement of the first several ammonium ions

Our team of scientists has experience in all areas of research including Life Science, Material Science, Chemical Synthesis, Chromatography, Analytical and many others.

Contact Technical Service