Skip to Content
Merck
All Photos(3)

Key Documents

455571

Sigma-Aldrich

Potassium borohydride

≥97%

Synonym(s):

Potassium tetrahydroborate

Sign Into View Organizational & Contract Pricing


About This Item

Linear Formula:
KBH4
CAS Number:
Molecular Weight:
53.94
EC Number:
MDL number:
UNSPSC Code:
12352306
PubChem Substance ID:
NACRES:
NA.22
Assay:
≥97%
form:
powder

Quality Level

Assay

≥97%

form

powder

reaction suitability

reagent type: reductant

mp

500 °C (dec.) (lit.)

density

1.18 g/mL at 25 °C (lit.)

SMILES string

[K+].[H][B-]([H])([H])[H]

InChI

1S/BH4.K/h1H4;/q-1;+1

InChI key

ICRGAIPBTSPUEX-UHFFFAOYSA-N

Looking for similar products? Visit Product Comparison Guide

Application

Potassium borohydride (KBH4) can be used as a reducing agent for the synthesis of:       
  • Copper nanoparticles by the reduction of copper salts.
  •  Enantioselective allylic alcohols by the catalytic reduction of enones and ketones in the presence of chiral N,N′-dioxidescandium(III) complex catalyst.,·        
  • Azoxybenzenes by the reduction of nitroarenes in the presence of phase transfer catalyst PEG-400.

It can also be employed as a boron source in the nickel–boron alloy, which is electrodeposited on carbon fiber to improve catalytic graphitization of carbon.

Signal Word

Danger

Hazard Statements

Hazard Classifications

Acute Tox. 3 Oral - Eye Dam. 1 - Repr. 1B - Skin Corr. 1B - Water-react 1

Storage Class Code

4.3 - Hazardous materials which set free flammable gases upon contact with water

WGK

nwg

Flash Point(F)

Not applicable

Flash Point(C)

Not applicable


Choose from one of the most recent versions:

Certificates of Analysis (COA)

Lot/Batch Number

Don't see the Right Version?

If you require a particular version, you can look up a specific certificate by the Lot or Batch number.

Already Own This Product?

Find documentation for the products that you have recently purchased in the Document Library.

Visit the Document Library

Preparation of copper nanoparticles by chemical reduction method using potassium borohydride
Zhang Q-L, et al.
Transactions of Nonferrous Metals Society of China, 20, 240-s244-240-s244 (2010)
Catalytic graphitization of carbon fibers with electrodeposited Ni-B alloy coating
Zhou H, et al.
Materials Chemistry and Physics, 110(2-3), 434-439 (2008)
S. Yan and S. Zhang,
International Journal of Hydrogen Energy, 36, 13392-13397 (2011)
A. Yin, et al.,
Applied Catalysis, 400, 39-47 (2011)
Gang Lian et al.
Dalton transactions (Cambridge, England : 2003), 40(26), 6961-6967 (2011-06-07)
Cubic boron nitride (cBN) is synthesized by a low-temperature solid state synthesis and in situ phase transformation route with NH(4)BF(4), B, NaBH(4) and KBH(4) as the boron sources and NaN(3) as the nitrogen source. Furthermore, two new strategies are developed

Our team of scientists has experience in all areas of research including Life Science, Material Science, Chemical Synthesis, Chromatography, Analytical and many others.

Contact Technical Service