Skip to Content
Merck
All Photos(1)

Key Documents

455202

Sigma-Aldrich

Tetrakis(diethylamido)hafnium(IV)

99.99%

Synonym(s):

Tetrakis(diethylamino)hafnium(IV)

Sign Into View Organizational & Contract Pricing


About This Item

Linear Formula:
[(CH2CH3)2N]4Hf
CAS Number:
Molecular Weight:
467.01
MDL number:
UNSPSC Code:
12352202
PubChem Substance ID:
NACRES:
NA.23

Assay

99.99%

form

liquid

reaction suitability

core: hafnium

bp

130 °C/0.01 mmHg (lit.)

density

1.249 g/mL at 25 °C (lit.)

SMILES string

CCN(CC)[Hf](N(CC)CC)(N(CC)CC)N(CC)CC

InChI

1S/4C4H10N.Hf/c4*1-3-5-4-2;/h4*3-4H2,1-2H3;/q4*-1;+4

InChI key

VBCSQFQVDXIOJL-UHFFFAOYSA-N

Looking for similar products? Visit Product Comparison Guide

General description

Atomic number of base material: 72 Hafnium

Analysis Note

Purity excludes ~2000ppm zirconium.

Pictograms

FlameExclamation mark

Signal Word

Danger

Hazard Statements

Hazard Classifications

Eye Irrit. 2 - Flam. Liq. 2 - Skin Irrit. 2 - STOT SE 3 - Water-react 2

Target Organs

Respiratory system

Supplementary Hazards

Storage Class Code

4.3 - Hazardous materials which set free flammable gases upon contact with water

WGK

WGK 3

Flash Point(F)

50.0 °F - closed cup

Flash Point(C)

10 °C - closed cup

Personal Protective Equipment

dust mask type N95 (US), Eyeshields, Gloves

Choose from one of the most recent versions:

Certificates of Analysis (COA)

Lot/Batch Number

Don't see the Right Version?

If you require a particular version, you can look up a specific certificate by the Lot or Batch number.

Already Own This Product?

Find documentation for the products that you have recently purchased in the Document Library.

Visit the Document Library

Articles

Thin film photovoltaic devices have become increasingly important in efficiently harnessing solar energy to meet consumer demand.

Our team of scientists has experience in all areas of research including Life Science, Material Science, Chemical Synthesis, Chromatography, Analytical and many others.

Contact Technical Service