Skip to Content
Merck
All Photos(1)

Key Documents

454982

Sigma-Aldrich

3-(Acryloyloxy)-2-hydroxypropyl methacrylate

Synonym(s):

1-(Acryloyloxy)-2-hydroxy-3-(methacryloyloxy)propane, 2-Hydroxy-1-acryloxy-3-methacryloxypropane, 2-Hydroxy-1-acryloyloxy-3-methacryloyloxypropane, 3-Acryloyloxy glycerol monomethacrylate

Sign Into View Organizational & Contract Pricing


About This Item

Linear Formula:
H2C=C(CH3)CO2CH2CH(OH)CH2O2CCH=CH2
CAS Number:
Molecular Weight:
214.22
MDL number:
UNSPSC Code:
12162002
PubChem Substance ID:
NACRES:
NA.23

contains

200 ppm monomethyl ether hydroquinone as inhibitor
350 ppm BHT as inhibitor

Quality Level

refractive index

n20/D 1.473 (lit.)

density

1.14 g/mL at 25 °C (lit.)

storage temp.

2-8°C

SMILES string

CC(=C)C(=O)OCC(O)COC(=O)C=C

InChI

1S/C10H14O5/c1-4-9(12)14-5-8(11)6-15-10(13)7(2)3/h4,8,11H,1-2,5-6H2,3H3

InChI key

ZODNDDPVCIAZIQ-UHFFFAOYSA-N

Application

  • Spectroscopic Analysis on Michael Addition Reaction of Secondary Amino Groups on Silica Surface with 3-(Acryloyloxy)-2-hydroxypropyl Methacrylate: This study discusses the introduction of polymerizable methacrylate groups to silica surfaces via Michael addition of AHM, observing increased functional group introduction with increased molar ratios (S Lee, KR Ha, 2014).
  • Synthesis of well-defined diblock copolymer nano-objects by RAFT non-aqueous emulsion polymerization: The paper elaborates on the synthesis of well-defined polymeric nano-objects employing RAFT polymerization in non-aqueous media, highlighting the versatility of methacrylate copolymers in creating tailored nanostructures (RR Gibson, A Fernyhough, OM Musa, SP Armes, 2021).
  • Spectroscopic analysis of methacrylate groups introduced on silica particle surfaces by the aza-Michael addition reaction: This publication explores the functionalization of silica particles with methacrylate groups using AHM, focusing on the efficiency of grafting via spectroscopic analysis (S Lee, KR Ha, 2016).
  • Antimicrobial and Responsive Zwitterionic Polymer Based on Cysteine Methacrylate Synthesized via RAFT Polymerization: Discusses the synthesis of a responsive, antimicrobial zwitterionic polymer from cysteine and AHM, using thiol-Michael reaction for antimicrobial applications (P Yadav, S Hafeez, J Jaishankar, P Srivastava, 2021).

Pictograms

CorrosionExclamation mark

Signal Word

Danger

Hazard Statements

Hazard Classifications

Eye Dam. 1 - Skin Irrit. 2 - Skin Sens. 1

Storage Class Code

10 - Combustible liquids

WGK

WGK 3

Flash Point(F)

235.4 °F - closed cup

Flash Point(C)

113 °C - closed cup

Personal Protective Equipment

dust mask type N95 (US), Eyeshields, Gloves

Certificates of Analysis (COA)

Search for Certificates of Analysis (COA) by entering the products Lot/Batch Number. Lot and Batch Numbers can be found on a product’s label following the words ‘Lot’ or ‘Batch’.

Already Own This Product?

Find documentation for the products that you have recently purchased in the Document Library.

Visit the Document Library

M Ugurlu
Australian dental journal, 65(3), 181-188 (2020-01-05)
This study aimed to evaluate the effect of double application of universal adhesives on bond strength to dentine after ionizing radiation. One hundred and forty caries-free human third molars were used. Half of the teeth were irradiated (IR) (70.2 Gy). The
Grzegorz Sokolowski et al.
Materials (Basel, Switzerland), 11(6) (2018-06-13)
Resin matrix dental materials undergo contraction and expansion changes due to polymerization and water absorption. Both phenomena deform resin-dentin bonding and influence the stress state in restored tooth structure in two opposite directions. The study tested three composite resin cements
San San May Phyo Aung et al.
Journal of dentistry, 104, 103527-103527 (2020-11-15)
Nowadays, the universal adhesives are often used for silane application prior to application of self-adhesive resin cements. The purpose of this study was to evaluate enamel surface roughness and to observe the enamel-adhesive interface after acid-base challenge using three different
Chenmin Yao et al.
The journal of adhesive dentistry, 22(1), 65-77 (2020-02-08)
This study investigated the bonding efficacy of a new so-called self-adhesive composite hybrid onto flat (FLAT) and high C-factor class-I cavity-bottom (CAVITY) dentin. The immediate and aged (50,000 thermocycles) microtensile bond strength (μTBS) to FLAT and CAVITY dentin of the
John Jackson Yang et al.
Microorganisms, 8(2) (2020-01-25)
Methicillin-resistant Staphylococcus aureus (S. aureus) (MRSA) has become an alarming threat to public health, and infected soft tissue. Antibiotics are commonly used to treat skin infection with MRSA, but the inappropriate use of antibiotics runs a considerable risk of generating

Our team of scientists has experience in all areas of research including Life Science, Material Science, Chemical Synthesis, Chromatography, Analytical and many others.

Contact Technical Service