Skip to Content
Merck
All Photos(1)

Key Documents

454818

Sigma-Aldrich

Diheptyl phthalate

97%

Synonym(s):

Phthalic acid diheptyl ester

Sign Into View Organizational & Contract Pricing


About This Item

Linear Formula:
C6H4-1,2-[CO2(CH2)6CH3]2
CAS Number:
Molecular Weight:
362.50
Beilstein:
2336623
EC Number:
MDL number:
UNSPSC Code:
12162002
PubChem Substance ID:
NACRES:
NA.23

Quality Level

Assay

97%

refractive index

n20/D 1.485 (lit.)

bp

195 °C/0.04 mmHg (lit.)

density

0.988 g/mL at 25 °C (lit.)

SMILES string

CCCCCCCOC(=O)c1ccccc1C(=O)OCCCCCCC

InChI

1S/C22H34O4/c1-3-5-7-9-13-17-25-21(23)19-15-11-12-16-20(19)22(24)26-18-14-10-8-6-4-2/h11-12,15-16H,3-10,13-14,17-18H2,1-2H3

InChI key

JQCXWCOOWVGKMT-UHFFFAOYSA-N

Looking for similar products? Visit Product Comparison Guide

Pictograms

Health hazardExclamation mark

Signal Word

Warning

Hazard Statements

Hazard Classifications

Eye Irrit. 2 - Repr. 2 - Skin Irrit. 2 - STOT SE 3

Target Organs

Respiratory system

Storage Class Code

10 - Combustible liquids

WGK

WGK 3

Flash Point(F)

235.4 °F - closed cup

Flash Point(C)

113 °C - closed cup

Personal Protective Equipment

dust mask type N95 (US), Eyeshields, Gloves

Choose from one of the most recent versions:

Certificates of Analysis (COA)

Lot/Batch Number

Don't see the Right Version?

If you require a particular version, you can look up a specific certificate by the Lot or Batch number.

Already Own This Product?

Find documentation for the products that you have recently purchased in the Document Library.

Visit the Document Library

Y Matsushima et al.
Eisei Shikenjo hokoku. Bulletin of National Institute of Hygienic Sciences, (110)(110), 26-31 (1992-01-01)
A twenty-eight day repeated oral dose toxicity test of dihepthyl phthalate (DHP) was carried out in male and female F344 rats at the dose levels of 0, 0.2, 1 and 5 g/kg/day. All rats in each group, consisting of 5
Meilan Jin et al.
Toxicology, 264(1-2), 16-25 (2009-08-01)
To investigate possible potential inducing preneoplastic lesions in liver and in vivo genotoxic potential of diheptyl phthalate (DHP), male F344 rats were subjected to repeated oral administration of DHP at 0, 2.5 or 5 g/kg/day for 28 days. In addition
Richard H McKee et al.
Reproductive toxicology (Elmsford, N.Y.), 21(3), 241-252 (2005-10-27)
Di-isoheptyl phthalate (DIHP) is a branched, phthalate ester with seven carbon alkyl side chains. Since structurally similar phthalates have been shown to produce developmental and/or reproductive effects in rodents, the potential for DIHP to produce developmental and reproductive toxicity was
Fumiyuki Nakane et al.
The Journal of toxicological sciences, 37(3), 527-537 (2012-06-13)
The 26-week oral toxicity of diheptyl phthalate (DHP), a peroxisome proliferator-activated receptor α (PPARα) agonist, with special emphasis on the potential induction of hepatocellular proliferative lesions was investigated in this study. DHP was administered to male F344 rats via gavage
S M Jickells et al.
Food additives and contaminants, 22(8), 768-782 (2005-09-09)
The gas phase transfer of substances from carton board (CB) and corrugated box board (CBB) through intervening layers to foods was studied. Substances covering a boiling point range of 252-425 degrees C and a range of polarities were incorporated into

Our team of scientists has experience in all areas of research including Life Science, Material Science, Chemical Synthesis, Chromatography, Analytical and many others.

Contact Technical Service