Skip to Content
Merck
All Photos(1)

Key Documents

449792

Sigma-Aldrich

Erbium(III) chloride

anhydrous, powder, 99.9% trace metals basis

Synonym(s):

Erbium trichloride

Sign Into View Organizational & Contract Pricing


About This Item

Linear Formula:
ErCl3
CAS Number:
Molecular Weight:
273.62
EC Number:
MDL number:
UNSPSC Code:
12352302
PubChem Substance ID:
NACRES:
NA.23
Assay:
99.9% trace metals basis
grade:
anhydrous
form:
powder

grade

anhydrous

Quality Level

Assay

99.9% trace metals basis

form

powder

reaction suitability

reagent type: catalyst
core: erbium

impurities

≤1500.0 ppm Trace Rare Earth Analysis

mp

774 °C (lit.)

density

4.1 g/mL at 25 °C (lit.)

SMILES string

Cl[Er](Cl)Cl

InChI

1S/3ClH.Er/h3*1H;/q;;;+3/p-3

InChI key

HDGGAKOVUDZYES-UHFFFAOYSA-K

Looking for similar products? Visit Product Comparison Guide

Application

Starting material for new edge-bridged octahedral M6 cluster compounds such as CsErTa6Cl18, for which electron and stability studies were performed.

Pictograms

Exclamation mark

Signal Word

Warning

Hazard Statements

Hazard Classifications

Eye Irrit. 2 - Skin Irrit. 2 - STOT SE 3

Target Organs

Respiratory system

Storage Class Code

11 - Combustible Solids

WGK

WGK 2

Flash Point(F)

Not applicable

Flash Point(C)

Not applicable

Personal Protective Equipment

dust mask type N95 (US), Eyeshields, Gloves

Choose from one of the most recent versions:

Certificates of Analysis (COA)

Lot/Batch Number

Don't see the Right Version?

If you require a particular version, you can look up a specific certificate by the Lot or Batch number.

Already Own This Product?

Find documentation for the products that you have recently purchased in the Document Library.

Visit the Document Library

A P Russell et al.
Analytical biochemistry, 164(1), 181-189 (1987-07-01)
A method of measuring the volumetric magnetic susceptibility, in which magnetically labeled cells or other particles are suspended in a paramagnetic solution of known susceptibility over the poles of a magnet, is presented. If the cells are more magnetic than
M Zborowski et al.
Journal of general microbiology, 138(1), 63-68 (1992-01-01)
A trivalent lanthanide ion, erbium (Er3+), has been used in combination with a magnetic separation technique to isolate seven bacterial species from suspensions in 0.9% saline. Erbium has an exceptionally high atomic magnetic moment of 9.3 Bohr magnetons, and following
Heng Qin et al.
Physical chemistry chemical physics : PCCP, 19(29), 19159-19167 (2017-07-14)
Efficient enhancement of photoluminescence in rare-earth activated upconversion materials is of great significance for their practical applications in various fields. In this work, three-dimensional mesoporous gold films were fabricated by a low-cost and facile dealloying approach to improve the upconversion
Duc Tu Vu et al.
Biomedical optics express, 12(1), 1-19 (2021-03-05)
We introduce a compact array fluorescence sensor principle that takes advantage of the long luminescence lifetimes of upconversion nanoparticles (UCNPs) to deploy a filter-free, optics-less contact geometry, advantageous for modern biochemical assays of biomolecules, pollutants or cells. Based on technologically
Angshuman Ray Chowdhuri et al.
Dalton transactions (Cambridge, England : 2003), 45(45), 18120-18132 (2016-10-28)
In this work, a new theranostic nanoplatform is developed to construct an anticancer drug carrier by integrating the distinct advantages of upconversion nanoparticles (UCNPs) and metal organic frameworks (MOFs) encapsulated with a targeting ligand. Here NaYF

Articles

Rare earth elements are vital in everyday life worldwide: catalysts in cars, colors in screens, magnets in electronics. Essential for modern living.

Rare earth elements are vital in everyday life worldwide: catalysts in cars, colors in screens, magnets in electronics. Essential for modern living.

Rare earth elements are vital in everyday life worldwide: catalysts in cars, colors in screens, magnets in electronics. Essential for modern living.

Rare earth elements are vital in everyday life worldwide: catalysts in cars, colors in screens, magnets in electronics. Essential for modern living.

Our team of scientists has experience in all areas of research including Life Science, Material Science, Chemical Synthesis, Chromatography, Analytical and many others.

Contact Technical Service