Skip to Content
Merck
All Photos(1)

Key Documents

448249

Sigma-Aldrich

Stearic-d35 acid

98 atom % D, 99% (CP)

Synonym(s):

Octadecanoic-d35 acid

Sign Into View Organizational & Contract Pricing


About This Item

Linear Formula:
CD3(CD2)16CO2H
CAS Number:
Molecular Weight:
319.69
MDL number:
UNSPSC Code:
12352106
PubChem Substance ID:
NACRES:
NA.12

isotopic purity

98 atom % D

Quality Level

Assay

99% (CP)

form

solid

bp

361 °C (lit.)

mp

68-70 °C (lit.)

mass shift

M+35

SMILES string

[2H]C([2H])([2H])C([2H])([2H])C([2H])([2H])C([2H])([2H])C([2H])([2H])C([2H])([2H])C([2H])([2H])C([2H])([2H])C([2H])([2H])C([2H])([2H])C([2H])([2H])C([2H])([2H])C([2H])([2H])C([2H])([2H])C([2H])([2H])C([2H])([2H])C([2H])([2H])C(O)=O

InChI

1S/C18H36O2/c1-2-3-4-5-6-7-8-9-10-11-12-13-14-15-16-17-18(19)20/h2-17H2,1H3,(H,19,20)/i1D3,2D2,3D2,4D2,5D2,6D2,7D2,8D2,9D2,10D2,11D2,12D2,13D2,14D2,15D2,16D2,17D2

InChI key

QIQXTHQIDYTFRH-KNAXIHRDSA-N

Packaging

This product may be available from bulk stock and can be packaged on demand. For information on pricing, availability and packaging, please contact Stable Isotopes Customer Service.

Storage Class Code

11 - Combustible Solids

WGK

nwg

Flash Point(F)

235.4 °F - closed cup

Flash Point(C)

113.00 °C - closed cup


Certificates of Analysis (COA)

Search for Certificates of Analysis (COA) by entering the products Lot/Batch Number. Lot and Batch Numbers can be found on a product’s label following the words ‘Lot’ or ‘Batch’.

Already Own This Product?

Find documentation for the products that you have recently purchased in the Document Library.

Visit the Document Library

Barbora Amélie Čuříková-Kindlová et al.
International journal of pharmaceutics, 563, 384-394 (2019-04-09)
Ceramides (Cers) are significant constituents of the stratum corneum (SC), the uppermost skin layer responsible for skin barrier properties. Cers are a heterogeneous group of lipids whose mutual interactions are still unclear. To better understand these interactions, we characterized model
Qin Yang et al.
Lipids in health and disease, 16(1), 10-10 (2017-01-18)
Dietary polyunsaturated fatty acids (PUFAs), especially n-3 PUFAs, are important for human health. The intestinal tract, a location that is heavily colonized by microorganisms, is the main organ for absorbing fatty acids. The purpose of this study was to analyze
Juan P Rodríguez et al.
Cancers, 11(4) (2019-04-25)
Hydroxy fatty acids are known to cause cell cycle arrest and apoptosis. The best studied of them, 9-hydroxystearic acid (9-HSA), induces apoptosis in cell lines by acting through mechanisms involving different targets. Using mass spectrometry-based lipidomic approaches, we show in

Our team of scientists has experience in all areas of research including Life Science, Material Science, Chemical Synthesis, Chromatography, Analytical and many others.

Contact Technical Service