Recommended Products
Quality Level
Assay
99%
form
powder
mp
163-166 °C (lit.)
functional group
ketone
phenyl
SMILES string
O=C(c1ccccc1)c2ccc(cc2)C(=O)c3ccccc3
InChI
1S/C20H14O2/c21-19(15-7-3-1-4-8-15)17-11-13-18(14-12-17)20(22)16-9-5-2-6-10-16/h1-14H
InChI key
NPENBPVOAXERED-UHFFFAOYSA-N
Related Categories
Storage Class Code
11 - Combustible Solids
WGK
WGK 3
Flash Point(F)
Not applicable
Flash Point(C)
Not applicable
Personal Protective Equipment
dust mask type N95 (US), Eyeshields, Gloves
Choose from one of the most recent versions:
Already Own This Product?
Find documentation for the products that you have recently purchased in the Document Library.
The potential application of poly (1, 4-phenylene diphenylvinylene), p-PDV for oxygen detection based on fluorescence quenching.
Sensors and Actuators B, Chemical, 96(3), 537-540 (2003)
1,4-Dibenzoylbenzene.
Acta Crystallographica Section C, Crystal Structure Communications, 48(9), 1715-1717 (1992)
The journal of physical chemistry. B, 119(6), 2241-2252 (2014-08-21)
The formation mechanism of ketyl radicals and several other selective photoreactions of benzophenone and its derivatives are initiated by the protonation of their triplet state and have been investigated using nanosecond time-resolved resonance Raman spectroscopy (ns-TR(3)) in solutions of varying
Our team of scientists has experience in all areas of research including Life Science, Material Science, Chemical Synthesis, Chromatography, Analytical and many others.
Contact Technical Service