Skip to Content
Merck
All Photos(1)

Key Documents

444960

Sigma-Aldrich

Methyl-d3 methacrylate-d5

≥99 atom % D, ≥99% (CP), contains hydroquinone as stabilizer

Synonym(s):

Methyl methacrylate-d8

Sign Into View Organizational & Contract Pricing


About This Item

Linear Formula:
D2C=C(CD3)CO2CD3
CAS Number:
Molecular Weight:
108.17
MDL number:
UNSPSC Code:
12352002
PubChem Substance ID:
NACRES:
NA.12

isotopic purity

≥99 atom % D

Quality Level

Assay

≥99% (CP)

contains

hydroquinone as stabilizer

refractive index

n20/D 1.413 (lit.)

bp

100 °C (lit.)

mp

−48 °C (lit.)

density

1.011 g/mL at 25 °C

mass shift

M+8

storage temp.

2-8°C

SMILES string

[2H]\C([2H])=C(\C(=O)OC([2H])([2H])[2H])C([2H])([2H])[2H]

InChI

1S/C5H8O2/c1-4(2)5(6)7-3/h1H2,2-3H3/i1D2,2D3,3D3

InChI key

VVQNEPGJFQJSBK-MGGKGRBPSA-N

Packaging

This product may be available from bulk stock and can be packaged on demand. For information on pricing, availability and packaging, please contact Stable Isotopes Customer Service.

Pictograms

FlameExclamation mark

Signal Word

Danger

Hazard Statements

Hazard Classifications

Flam. Liq. 2 - Skin Irrit. 2 - Skin Sens. 1B - STOT SE 3

Target Organs

Respiratory system

Storage Class Code

3 - Flammable liquids

WGK

WGK 1

Flash Point(F)

48.2 °F - closed cup

Flash Point(C)

9 °C - closed cup


Choose from one of the most recent versions:

Certificates of Analysis (COA)

Lot/Batch Number

Don't see the Right Version?

If you require a particular version, you can look up a specific certificate by the Lot or Batch number.

Already Own This Product?

Find documentation for the products that you have recently purchased in the Document Library.

Visit the Document Library

Li Zhou et al.
The journal of physical chemistry. A, 121(23), 4464-4474 (2017-05-13)
Two different experimental methods (relative rate and absolute rate methods) were used to measure the rate coefficients for the reactions of NO

Our team of scientists has experience in all areas of research including Life Science, Material Science, Chemical Synthesis, Chromatography, Analytical and many others.

Contact Technical Service