Skip to Content
Merck
All Photos(1)

Key Documents

441252

Sigma-Aldrich

D-Pinitol

95%

Synonym(s):

3-O-Methyl-D-chiro-inositol

Sign Into View Organizational & Contract Pricing


About This Item

Empirical Formula (Hill Notation):
C7H14O6
CAS Number:
Molecular Weight:
194.18
MDL number:
UNSPSC Code:
12352112
PubChem Substance ID:
NACRES:
NA.22

Assay

95%

form

powder

optical activity

[α]20/D 60.0 to 70.0°, c = 1% in H2O

mp

179-185 °C (lit.)

functional group

ether
hydroxyl

SMILES string

CO[C@@H]1[C@@H](O)[C@@H](O)[C@H](O)[C@H](O)[C@H]1O

InChI

1S/C7H14O6/c1-13-7-5(11)3(9)2(8)4(10)6(7)12/h2-12H,1H3/t2-,3-,4-,5-,6+,7+/m0/s1

InChI key

DSCFFEYYQKSRSV-KLJZZCKASA-N

Looking for similar products? Visit Product Comparison Guide

General description

D-pinitol, commonly found conifers, is an isomer of L-quebrachitol.

Application

D-pinitol may be used as a starting material to prepare its azole nucleoside analogs. It may also be used in the preparation of 1D-1,5-dideoxy-1,5-difluoro-neo-inositol and 1D-1-deoxy-1-fluoro-myo-inositol.
Precursor to biologically active fluorinated isosteres of inositol, which show cell growth inhibitory properties. Has shown antidiabetic properties in mice. Believed to be a salt stress regulator in a wide range of plants.

Storage Class Code

11 - Combustible Solids

WGK

WGK 3

Flash Point(F)

Not applicable

Flash Point(C)

Not applicable

Personal Protective Equipment

dust mask type N95 (US), Eyeshields, Gloves

Choose from one of the most recent versions:

Certificates of Analysis (COA)

Lot/Batch Number

Don't see the Right Version?

If you require a particular version, you can look up a specific certificate by the Lot or Batch number.

Already Own This Product?

Find documentation for the products that you have recently purchased in the Document Library.

Visit the Document Library

Customers Also Viewed

Slide 1 of 1

1 of 1

Hyun Jin Kim et al.
Annals of nutrition & metabolism, 60(1), 1-5 (2011-12-20)
Pinitol is thought to mediate insulin action and improve insulin resistance. We evaluated the effects of pinitol on glycemic control, insulin resistance and adipocytokine levels in type 2 diabetic patients. A total of 66 patients with type 2 diabetes who
Narayanan, C.R. et al.
Current Science, 56, 139-139 (1987)
Binika Hada et al.
The journals of gerontology. Series A, Biological sciences and medical sciences, 68(3), 226-234 (2012-07-31)
D-chiro-inositol, a member of the inositol family, and pinitol, a 3-methoxy analogue of D-chiro-inositol, have been proposed to have antidiabetic, antiinflammatory, anticancer and stamina enhancing effects. We found that supplementing the diet of Drosophila with D-chiro-inositol and pinitol extended adult
Selvaraj Sivakumar et al.
General physiology and biophysics, 28(3), 233-241 (2009-12-29)
During diabetes mellitus, endogenous hepatic glucose production is increased as a result of impaired activities of the key enzymes of carbohydrate metabolism, which leads to the condition known as hyperglycemia. D-pinitol, a bioactive constituent isolated from soybeans, has been shown
Kozikowski, A.P. et al.
Journal of the American Chemical Society, 112, 4528-4528 (1990)

Our team of scientists has experience in all areas of research including Life Science, Material Science, Chemical Synthesis, Chromatography, Analytical and many others.

Contact Technical Service