Skip to Content
Merck
All Photos(4)

Key Documents

440752

Sigma-Aldrich

Polycaprolactone

average Mw ~14,000, average Mn ~10,000 by GPC

Synonym(s):

2-Oxepanone homopolymer, 6-Caprolactone polymer

Sign Into View Organizational & Contract Pricing


About This Item

Linear Formula:
(C6H10O2)n
UNSPSC Code:
12162002
NACRES:
NA.23

form

flakes or chunk(s)

Quality Level

mol wt

average Mn ~10,000 by GPC
average Mw ~14,000

viscosity

400-1000 mPa.s, 50 wt. % in xylene

mp

60 °C (lit.)

transition temp

Tg −60 °C

density

1.146 g/mL at 25 °C

Looking for similar products? Visit Product Comparison Guide

General description

Polycaprolactone is a α,ω-dihydroxy functional polymer.

Application

Polycaprolactone may find use as an extrusion aid, die lubricant, mold release, pigment and filler dispersion aid and polyester segments in urethanes and block polyesters.

Features and Benefits

Biodegradable polymer
Non-toxic, biodegradable in soil, broad miscibility, mechanical compatibility with many polymers and good adhesion to a broad spectrum of substrates.

Physical form

α,ω-dihydroxy functional polymer.

Storage Class Code

11 - Combustible Solids

WGK

WGK 3

Flash Point(F)

>230.0 °F

Flash Point(C)

> 110 °C

Personal Protective Equipment

dust mask type N95 (US), Eyeshields, Gloves

Certificates of Analysis (COA)

Search for Certificates of Analysis (COA) by entering the products Lot/Batch Number. Lot and Batch Numbers can be found on a product’s label following the words ‘Lot’ or ‘Batch’.

Already Own This Product?

Find documentation for the products that you have recently purchased in the Document Library.

Visit the Document Library

Customers Also Viewed

Slide 1 of 1

1 of 1

Kyungjin Park et al.
Scientific reports, 7(1), 13359-13359 (2017-10-19)
Optical resolution photoacoustic microscopy (OR-PAM) is a non-invasive, label-free method of in vivo imaging with microscopic resolution and high optical contrast. Based on intrinsic contrasts, OR-PAM has expanded to include in vivo vessel imaging, flow cytometry, physiological parameter analysis, and
Jack R T Darby et al.
The Journal of physiology, 597(20), 5063-5077 (2019-09-05)
Substrate restriction during critical developmental windows of gestation programmes offspring for a predisposition towards cardiovascular disease in adult life. This study aimed to determine the effect of maternal resveratrol (RSV) treatment in an animal model in which chronic fetal catheterisation
Jixin Xue et al.
Biomaterials, 34(11), 2624-2631 (2013-01-29)
Tissue engineering approach continuously requires for emerging strategies to improve the efficacy in repairing and regeneration of tissue defects. Previously, we developed a sandwich model strategy for cartilage engineering, using the combination of acellular cartilage sheets (ACSs) and chondrocytes. However
P R Sreerekha et al.
Journal of biomedical nanotechnology, 9(5), 790-800 (2013-06-28)
Fabricating scaffolds mimicking the native extracellular matrix (ECM) in both structure and function is a key challenge in the field of tissue engineering. Previously we have demonstrated a novel electrospinnig method for the fabrication of fibrin nanofibers using Poly(vinyl alcohol)
Kalaipriya Madhaiyan et al.
International journal of pharmaceutics, 444(1-2), 70-76 (2013-02-02)
Biocompatible PCL polymer nanofiber mediated sustained release of hydrophilic drug and applicability as transdermal delivery system is attempted. This new attempt to investigate water soluble vitamin delivery with hydrophobic polymer nanofiber sustained the release of the vitamin and the method

Articles

In the past two decades, tissue engineering and regenerative medicine have become important interdisciplinary fields that span biology, chemistry, engineering, and medicine.

Innovations in polymer technology have had a significant impact on the advancement of novel drug delivery systems.

Our team of scientists has experience in all areas of research including Life Science, Material Science, Chemical Synthesis, Chromatography, Analytical and many others.

Contact Technical Service