Skip to Content
Merck
All Photos(2)

Key Documents

435139

Sigma-Aldrich

Poly(Bisphenol A carbonate)

melt index 10-12 g/10 min (300°C/1.2kg)

Synonym(s):

2,2-Bis(4-hydroxyphenyl)propane-carbonic acid copolymer, Bisphenol A carbonate polymer, Bisphenol A polycarbonate, Poly(bisphenol A carbonate), Poly(dian carbonate)

Sign Into View Organizational & Contract Pricing


About This Item

CAS Number:
MDL number:
UNSPSC Code:
12162002
PubChem Substance ID:
NACRES:
NA.23

form

pellets

Quality Level

melt index

10-12 g/10 min (300°C/1.2kg)

impact strength

801 J/m (Izod, ASTM D 256, 22.7 °C)

dielectric constant

2.96, 1 MHz (ASTM D 150)

hardness

118 (Rockwell R, ASTM D 785)

refractive index

n20/D 1.586

transition temp

softening point 154 °C (Vicat, rate B, ASTM D 1525)

density

1.2 g/mL at 25 °C (lit.)

SMILES string

CC(C)(c1ccc(O)cc1)c2ccc(O)cc2

InChI

1S/C15H16O2.CH2O3/c1-15(2,11-3-7-13(16)8-4-11)12-5-9-14(17)10-6-12;2-1(3)4/h3-10,16-17H,1-2H3;(H2,2,3,4)

InChI key

XSXWYGABGYBZRM-UHFFFAOYSA-N

Gene Information

mouse ... Esr1(13982)
rat ... Ar(24208)

Looking for similar products? Visit Product Comparison Guide

General description

Service temperature, max.135°C

Storage Class Code

11 - Combustible Solids

WGK

WGK 3

Flash Point(F)

Not applicable

Flash Point(C)

Not applicable

Personal Protective Equipment

dust mask type N95 (US), Eyeshields, Gloves

Choose from one of the most recent versions:

Certificates of Analysis (COA)

Lot/Batch Number

Don't see the Right Version?

If you require a particular version, you can look up a specific certificate by the Lot or Batch number.

Already Own This Product?

Find documentation for the products that you have recently purchased in the Document Library.

Visit the Document Library

Brian T Schaefer et al.
Nature communications, 11(1), 4163-4163 (2020-08-21)
Solid-state magnetic field sensors are important for applications in commercial electronics and fundamental materials research. Most magnetic field sensors function in a limited range of temperature and magnetic field, but Hall sensors in principle operate over a broad range of
Yuki Kitahara et al.
Chemosphere, 80(11), 1281-1284 (2010-07-16)
The thermal decomposition of poly(bisphenol A carbonate) (PoC) results in the formation of the endocrine disruptor bisphenol A (BPA). In the present work, we investigated the kinetics of the thermal decomposition of PoC, and the subsequent decomposition of BPA, under
Hiroshi Yabu et al.
Langmuir : the ACS journal of surfaces and colloids, 22(11), 4992-4997 (2006-05-17)
We describe here a preparation of pincushion structures with holes, hexagonally arranged microrings, and microdots by simple heating and peeling of self-organized honeycomb-patterned films. We have reported that the honeycomb-patterned films can be prepared by casting the solution of an
Vianney Souplet et al.
Methods in molecular biology (Clifton, N.J.), 570, 287-297 (2009-08-04)
Microarrays are frequently prepared on microscope glass slides. However, glass substrates can break or cut and thus can lead to the contamination of the manipulator during the analysis of biological samples. Alternately, bisphenol A polycarbonate (PC) is shock-resistant and, in
Molla R Islam et al.
Chemistry (Weinheim an der Bergstrasse, Germany), 17(22), 6098-6108 (2011-04-19)
Spontaneous phase-separated, controlled aggregate structures of photo- and electroactive molecules in polymer matrices are of interest for device fabrication. We show that the self-assembly of octabutoxyphthalocyanine (Pc) in polymer matrices leads to tubular morphology of Pc when the film is

Our team of scientists has experience in all areas of research including Life Science, Material Science, Chemical Synthesis, Chromatography, Analytical and many others.

Contact Technical Service