Skip to Content
Merck
All Photos(4)

Key Documents

434531

Sigma-Aldrich

Acetone-d6

99.9 atom % D, contains 0.03 % (v/v) TMS

Synonym(s):

Hexadeuteroacetone

Sign Into View Organizational & Contract Pricing


About This Item

Linear Formula:
CD3COCD3
CAS Number:
Molecular Weight:
64.12
Beilstein:
1702935
EC Number:
MDL number:
UNSPSC Code:
12142201
PubChem Substance ID:
NACRES:
NA.21

vapor density

2 (vs air)

Quality Level

vapor pressure

14.39 psi ( 55 °C)
184 mmHg ( 20 °C)
3.67 psi ( 20 °C)

isotopic purity

99.9 atom % D

Assay

≥99% (CP)

form

liquid

contains

0.03 % (v/v) TMS

expl. lim.

13.2 %

packaging

ampule of 1 mL, 10 g
glass bottle of 10 g, 25 g, 50 g

technique(s)

NMR: suitable

impurities

≤0.0500% water
water

refractive index

n20/D 1.355 (lit.)

bp

55.5 °C (lit.)

mp

−93.8 °C (lit.)

density

0.872 g/mL at 25 °C (lit.)

mass shift

M+6

SMILES string

[2H]C([2H])([2H])C(=O)C([2H])([2H])[2H]

InChI

1S/C3H6O/c1-3(2)4/h1-2H3/i1D3,2D3

InChI key

CSCPPACGZOOCGX-WFGJKAKNSA-N

Looking for similar products? Visit Product Comparison Guide

General description

Acetone-d6 is a deuterated NMR solvent containing 0.03% (v/v) TMS (tetramethylsilane). It is useful in NMR-based research and analyses. Its Infrared and Raman spectra have been analyzed. Pyrolysis and photolysis of acetone-d6 and methane mixtures have been investigated as a function of temperature. It has been reported to be prepared by reacting heavy water with propyne-d4 in the presence of mercury catalyst.

Application

Acetone-d6 may be used as an internal standard to detect aldehydes and acetone in water by headspace-solid-phase microextraction and gas chromatography-mass spectrometry.

Recommended products

Check out ChemisTwin®, our brand new online portal for identity confirmation and quantification of NMR spectra. Learn more or reach out to us for a free trial.

Pictograms

FlameExclamation mark

Signal Word

Danger

Hazard Statements

Hazard Classifications

Eye Irrit. 2 - Flam. Liq. 2 - STOT SE 3

Target Organs

Central nervous system

Supplementary Hazards

Storage Class Code

3 - Flammable liquids

WGK

WGK 1

Flash Point(F)

-2.2 °F - closed cup

Flash Point(C)

-19 °C - closed cup

Personal Protective Equipment

dust mask type N95 (US), Eyeshields, Gloves

Certificates of Analysis (COA)

Search for Certificates of Analysis (COA) by entering the products Lot/Batch Number. Lot and Batch Numbers can be found on a product’s label following the words ‘Lot’ or ‘Batch’.

Already Own This Product?

Find documentation for the products that you have recently purchased in the Document Library.

Visit the Document Library

Customers Also Viewed

Slide 1 of 1

1 of 1

Hyun-Ji Kim et al.
Journal of separation science, 34(6), 693-699 (2011-02-15)
We describe a simple and automatic method to determine nine aldehydes and acetone simultaneously in water. This method is based on derivatization with 2,2,2-trifluoroethylhydrazine (TFEH) and consecutive headspace-solid-phase microextraction and gas chromatography-mass spectrometry. Acetone-d(6) was used as the internal standard.
The Pyrolysis and Photolysis of Acetone-d6 in the Presence of Methane.
McNesby JR and Gordon AS.
Journal of the American Chemical Society, 76(16), 4196-4198 (1954)
Vibrational spectra and assignment of acetone, ααα acetone-d3 and acetone-d6.
Dellepiane G and Overend J.
Spectrochimica Acta Part A: Molecular Spectroscopy, 22(4), 593-614 (1966)
Neue Synthese von Aceton-d6.
Hunziker H, et al.
Helvetica Chimica Acta, 45(1), 59-62 (1962)
Eric Busseron et al.
ACS nano, 9(3), 2760-2772 (2015-03-04)
A family of triarylamine-fullerene conjugates has been synthesized and shown to self-assemble upon light stimulation in chlorinated solvents. This light-induced process primarily involves excitation of triarylamine derivatives, which then oxidize and stack with their neutral counterparts to form charge transfer

Our team of scientists has experience in all areas of research including Life Science, Material Science, Chemical Synthesis, Chromatography, Analytical and many others.

Contact Technical Service