Skip to Content
Merck
All Photos(1)

Key Documents

433624

Sigma-Aldrich

Magnesium peroxide complex

technical grade

Synonym(s):

Magnesium dioxide

Sign Into View Organizational & Contract Pricing


About This Item

Linear Formula:
MgO2 · xMgO
CAS Number:
EC Number:
UNSPSC Code:
12352303
PubChem Substance ID:
NACRES:
NA.23

grade

technical grade

Quality Level

form

powder

composition

Mg, 30%
MgO2, 24-28%

reaction suitability

reagent type: oxidant

application(s)

battery manufacturing

SMILES string

[Mg++].[O-][O-].O=[Mg]

InChI

1S/2Mg.O2.O/c;;1-2;/q;+2;-2;

InChI key

IODFUUWCSICZNX-UHFFFAOYSA-N

Pictograms

Flame over circle

Signal Word

Danger

Hazard Statements

Hazard Classifications

Ox. Sol. 2

Storage Class Code

5.1B - Oxidizing hazardous materials

WGK

WGK 3

Flash Point(F)

Not applicable

Flash Point(C)

Not applicable

Personal Protective Equipment

dust mask type N95 (US), Eyeshields, Gloves

Choose from one of the most recent versions:

Certificates of Analysis (COA)

Lot/Batch Number

Sorry, we don't have COAs for this product available online at this time.

If you need assistance, please contact Customer Support.

Already Own This Product?

Find documentation for the products that you have recently purchased in the Document Library.

Visit the Document Library

Our team of scientists has experience in all areas of research including Life Science, Material Science, Chemical Synthesis, Chromatography, Analytical and many others.

Contact Technical Service