Skip to Content
Merck
All Photos(1)

Key Documents

429406

Sigma-Aldrich

Cerium(III) chloride

AnhydroBeads, −10 mesh, ≥99.99% trace metals basis

Synonym(s):

Cerium trichloride, Cerous chloride

Sign Into View Organizational & Contract Pricing


About This Item

Empirical Formula (Hill Notation):
CeCl3
CAS Number:
Molecular Weight:
246.48
EC Number:
MDL number:
UNSPSC Code:
12352302
PubChem Substance ID:
NACRES:
NA.23
Assay:
≥99.99% trace metals basis
form:
beads
Pricing and availability is not currently available.

product line

AnhydroBeads

Quality Level

Assay

≥99.99% trace metals basis

form

beads

reaction suitability

reagent type: catalyst
core: cerium

impurities

≤100.0 ppm Trace Rare Earth Analysis

particle size

−10 mesh

mp

848 °C (lit.)

density

3.97 g/mL at 25 °C (lit.)

SMILES string

Cl[Ce](Cl)Cl

InChI

1S/Ce.3ClH/h;3*1H/q+3;;;/p-3

InChI key

VYLVYHXQOHJDJL-UHFFFAOYSA-K

Looking for similar products? Visit Product Comparison Guide

General description

Cerium(III) chloride is a non-toxic, water-tolerant, Lewis acid catalyst that imparts high regio-and chemoselectivity to various chemical transformations.[1] It is widely used as an environmentally friendly acidic promoter in many synthetic organic reactions such as C-C bond formation reaction, transition metal catalyze dhydrogenation, and reduction of C-O double bond.[2]

Application

  • The role of dynamic ligand exchange in the oxidation chemistry of cerium (III): This study explores the redox chemistry of cerium, utilizing Cerium(III) chloride in oxidation reactions to understand its potential in various chemical transformations (JR Robinson et al., 2016).
  • Photocatalytic C- C bond cleavage and amination of cycloalkanols by cerium (III) chloride complex: Examines the photocatalytic properties of a cerium(III) chloride complex, used for C-C bond cleavage and amination, highlighting its potential in organic synthesis (JJ Guo et al., 2016).
  • Dehydration, hydrolysis and oxidation of cerium chloride heptahydrate in air atmosphere: Investigates the behavior of hydrated forms of Cerium(III) chloride under varying conditions, contributing to understanding its stability and reactivity (S Xue et al., 2017).

Other Notes

Scoring knife is required to open the ampoule

Legal Information

AnhydroBeads is a trademark of Sigma-Aldrich Co. LLC

Pictograms

CorrosionEnvironment

Signal Word

Danger

Hazard Statements

Hazard Classifications

Aquatic Acute 1 - Aquatic Chronic 1 - Eye Dam. 1 - Skin Corr. 1C

Storage Class Code

8B - Non-combustible corrosive hazardous materials

WGK

WGK 2

Personal Protective Equipment

dust mask type N95 (US), Eyeshields, Gloves

Choose from one of the most recent versions:

Certificates of Analysis (COA)

Lot/Batch Number

Don't see the Right Version?

If you require a particular version, you can look up a specific certificate by the Lot or Batch number.

Already Own This Product?

Find documentation for the products that you have recently purchased in the Document Library.

Visit the Document Library

The use of anhydrous CeCl3 as a catalyst for the synthesis of 3-sulfenyl indoles
Claudio C. Silveira, et al.
Tetrahedron Letters, 51, 2014-2016 (2010)
Applications of CeCl3 as an Environmental Friendly Promoter in Organic Chemistry
Giuseppe Bartoli, et al.
Chemical Reviews, 110, 6104-6143 (2010)
Xiao, Z. Timberlake, J.W.
Tetrahedron, 54, 4211-4211 (1998)
Na Li et al.
Environmental toxicology, 28(6), 349-358 (2011-06-10)
To investigate the molecular mechanism of inflammatory response in the mouse liver caused by exposure to CeCl₃, we measured the liver indices, and cerium content, evaluated the liver histopathological section, detected serum biochemical parameters of liver function, and the immunoglobulin
Xiaoming Tao et al.
Organic letters, 15(1), 72-75 (2012-12-25)
Dynamic kinetic resolution of various α-amido-β-keto phosphonates via asymmetric hydrogenation proceeded efficiently to give the corresponding β-hydroxy-α-amido phosphonates in high diastereo- and enantioselectivities (up to 99:1 syn/anti, 99.8% ee). The addition of catalytic amounts of CeCl(3)·7H(2)O is necessary to achieve

Questions

Reviews

No rating value

Active Filters

Our team of scientists has experience in all areas of research including Life Science, Material Science, Chemical Synthesis, Chromatography, Analytical and many others.

Contact Technical Service